Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 128120291-128120564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UGGT1
Entrez Gene ID
56886
Gene Name
UDP-glucose glycoprotein glucosyltransferase 1
Gene Aliases
HUGT1, UGCGL1, UGT1
UniGene
Hs.743306
Gene Chromosome Location
Chr.2: 128091180-128195677 on Build GRCh38
Transcripts
NM_020120.3 NR_027671.2 XM_006712634.3 XM_006712635.3 XM_006712636.3 XM_017004507.1 XM_017004508.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1006375 c.895G>A p.G299R Chr.2: 128120378-128120378 on Build GRCh38
1006374 c.886G>A p.D296N Chr.2: 128120369-128120369 on Build GRCh38
1006373 c.885C>T p.P295P Chr.2: 128120368-128120368 on Build GRCh38
475892 c.963G>A p.W321* Chr.2: 128120446-128120446 on Build GRCh38
3113741 c.932G>A p.S311N Chr.2: 128120415-128120415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022847087 Chr.2: 128120382-128120382 on Build GRCh38
rs778451206 Chr.2: 128120407-128120407 on Build GRCh38
rs895536060 Chr.2: 128120368-128120368 on Build GRCh38
rs781085887 Chr.2: 128120498-128120499 on Build GRCh38
rs557524326 Chr.2: 128120333-128120333 on Build GRCh38
rs775442369 Chr.2: 128120432-128120432 on Build GRCh38
rs761002944 Chr.2: 128120343-128120343 on Build GRCh38
rs753974200 Chr.2: 128120357-128120357 on Build GRCh38
rs766803627 Chr.2: 128120479-128120479 on Build GRCh38
rs765273176 Chr.2: 128120454-128120454 on Build GRCh38
rs765347260 Chr.2: 128120369-128120369 on Build GRCh38
rs954162743 Chr.2: 128120519-128120519 on Build GRCh38
rs914720472 Chr.2: 128120512-128120512 on Build GRCh38
rs186851172 Chr.2: 128120470-128120470 on Build GRCh38
rs545017038 Chr.2: 128120518-128120518 on Build GRCh38
rs1026215962 Chr.2: 128120399-128120399 on Build GRCh38
rs80160667 Chr.2: 128120492-128120492 on Build GRCh38
rs915433423 Chr.2: 128120523-128120524 on Build GRCh38
rs763382463 Chr.2: 128120478-128120478 on Build GRCh38
rs537419232 Chr.2: 128120323-128120323 on Build GRCh38
rs1015484091 Chr.2: 128120438-128120438 on Build GRCh38
rs574148141 Chr.2: 128120437-128120437 on Build GRCh38
rs368495974 Chr.2: 128120480-128120480 on Build GRCh38
rs748922668 Chr.2: 128120393-128120393 on Build GRCh38
rs758153327 Chr.2: 128120528-128120528 on Build GRCh38
rs771807298 Chr.2: 128120324-128120324 on Build GRCh38
rs770392849 Chr.2: 128120396-128120396 on Build GRCh38
rs758897627 Chr.2: 128120372-128120372 on Build GRCh38
rs375538125 Chr.2: 128120315-128120315 on Build GRCh38
rs777052701 Chr.2: 128120443-128120443 on Build GRCh38
rs764356457 Chr.2: 128120354-128120354 on Build GRCh38
rs4662594 Chr.2: 128120440-128120440 on Build GRCh38
rs373739698 Chr.2: 128120332-128120332 on Build GRCh38
rs745827245 Chr.2: 128120420-128120420 on Build GRCh38
rs755391390 Chr.2: 128120505-128120505 on Build GRCh38
rs866870244 Chr.2: 128120447-128120447 on Build GRCh38
rs751862525 Chr.2: 128120381-128120381 on Build GRCh38
rs780304167 Chr.2: 128120377-128120377 on Build GRCh38
rs762053199 Chr.2: 128120359-128120359 on Build GRCh38
rs762074584 Chr.2: 128120449-128120449 on Build GRCh38
rs772000066 Chr.2: 128120425-128120425 on Build GRCh38
rs946891559 Chr.2: 128120525-128120525 on Build GRCh38
rs755766836 Chr.2: 128120388-128120388 on Build GRCh38
rs777466337 Chr.2: 128120391-128120391 on Build GRCh38

View Full Product Details