Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29673558-29673803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP57
Entrez Gene ID
346171
Gene Name
ZFP57 zinc finger protein
Gene Aliases
C6orf40, TNDM1, ZNF698, bA145L22, bA145L22.2
UniGene
Hs.156326
Gene Chromosome Location
Chr.6: 29672392-29681150 on Build GRCh38
Transcripts
NM_001109809.2 XM_006715087.3 XM_011514570.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
236621 c.319C>A p.Q107K Chr.6: 29673732-29673732 on Build GRCh38
5869990 c.420G>A p.M140I Chr.6: 29673631-29673631 on Build GRCh38
1621358 c.377G>T p.R126I Chr.6: 29673674-29673674 on Build GRCh38
1077282 c.335G>T p.R112I Chr.6: 29673716-29673716 on Build GRCh38
1642978 c.314G>A p.R105Q Chr.6: 29673737-29673737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778941830 Chr.6: 29673588-29673588 on Build GRCh38
rs752735462 Chr.6: 29673702-29673702 on Build GRCh38
rs754805449 Chr.6: 29673670-29673670 on Build GRCh38
rs749203567 Chr.6: 29673584-29673584 on Build GRCh38
rs971469760 Chr.6: 29673727-29673727 on Build GRCh38
rs765599364 Chr.6: 29673625-29673625 on Build GRCh38
rs757840663 Chr.6: 29673704-29673704 on Build GRCh38
rs185278475 Chr.6: 29673770-29673770 on Build GRCh38
rs777578657 Chr.6: 29673710-29673710 on Build GRCh38
rs751750269 Chr.6: 29673655-29673655 on Build GRCh38
rs746634665 Chr.6: 29673726-29673726 on Build GRCh38
rs772724131 Chr.6: 29673597-29673597 on Build GRCh38
rs747207653 Chr.6: 29673713-29673714 on Build GRCh38
rs542920653 Chr.6: 29673589-29673589 on Build GRCh38
rs200541066 Chr.6: 29673620-29673620 on Build GRCh38
rs964122823 Chr.6: 29673677-29673677 on Build GRCh38
rs762074022 Chr.6: 29673663-29673663 on Build GRCh38
rs778657283 Chr.6: 29673681-29673681 on Build GRCh38
rs754861035 Chr.6: 29673586-29673586 on Build GRCh38
rs764303925 Chr.6: 29673637-29673637 on Build GRCh38
rs968875761 Chr.6: 29673627-29673627 on Build GRCh38
rs983459674 Chr.6: 29673672-29673672 on Build GRCh38
rs922250508 Chr.6: 29673712-29673712 on Build GRCh38
rs748137597 Chr.6: 29673653-29673653 on Build GRCh38
rs1027550840 Chr.6: 29673652-29673652 on Build GRCh38
rs776081525 Chr.6: 29673628-29673628 on Build GRCh38
rs374760825 Chr.6: 29673724-29673724 on Build GRCh38
rs746180307 Chr.6: 29673767-29673767 on Build GRCh38
rs759998407 Chr.6: 29673621-29673621 on Build GRCh38
rs746724268 Chr.6: 29673607-29673607 on Build GRCh38
rs114591600 Chr.6: 29673737-29673737 on Build GRCh38
rs771827176 Chr.6: 29673654-29673654 on Build GRCh38
rs771387494 Chr.6: 29673616-29673616 on Build GRCh38
rs201703858 Chr.6: 29673636-29673636 on Build GRCh38
rs771624086 Chr.6: 29673593-29673593 on Build GRCh38
rs771073457 Chr.6: 29673762-29673762 on Build GRCh38
rs767749722 Chr.6: 29673667-29673667 on Build GRCh38

View Full Product Details