Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11722373-11722608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF823
Entrez Gene ID
55552
Gene Name
zinc finger protein 823
Gene Aliases
HSZFP36
UniGene
Hs.142167
Gene Chromosome Location
Chr.19: 11721265-11739009 on Build GRCh38
Transcripts
NM_001080493.3 NM_001297610.1 NM_017507.1 XM_017026941.1 XM_017026942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
564737 c.1042C>T p.R348* Chr.19: 11722492-11722492 on Build GRCh38
2814755 c.964C>T p.H322Y Chr.19: 11722570-11722570 on Build GRCh38
2814757 c.950C>A p.T317K Chr.19: 11722584-11722584 on Build GRCh38
5505956 c.1148_1149delCA p.T383fs*24 Chr.19: 11722385-11722386 on Build GRCh38
6833430 c.1107A>G p.L369L Chr.19: 11722427-11722427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759765537 Chr.19: 11722576-11722576 on Build GRCh38
rs756369831 Chr.19: 11722400-11722400 on Build GRCh38
rs753960639 Chr.19: 11722463-11722463 on Build GRCh38
rs749459878 Chr.19: 11722425-11722425 on Build GRCh38
rs932998150 Chr.19: 11722404-11722404 on Build GRCh38
rs767571818 Chr.19: 11722450-11722450 on Build GRCh38
rs779105933 Chr.19: 11722546-11722546 on Build GRCh38
rs779514791 Chr.19: 11722520-11722520 on Build GRCh38
rs957628011 Chr.19: 11722503-11722503 on Build GRCh38
rs200817135 Chr.19: 11722492-11722492 on Build GRCh38
rs773804721 Chr.19: 11722408-11722408 on Build GRCh38
rs976602797 Chr.19: 11722556-11722556 on Build GRCh38
rs372705042 Chr.19: 11722582-11722582 on Build GRCh38
rs199516132 Chr.19: 11722502-11722502 on Build GRCh38
rs756141328 Chr.19: 11722455-11722455 on Build GRCh38
rs751843264 Chr.19: 11722418-11722418 on Build GRCh38
rs746679475 Chr.19: 11722525-11722525 on Build GRCh38
rs144085286 Chr.19: 11722575-11722575 on Build GRCh38
rs761455162 Chr.19: 11722554-11722554 on Build GRCh38
rs771904079 Chr.19: 11722506-11722506 on Build GRCh38
rs763583081 Chr.19: 11722415-11722415 on Build GRCh38
rs780482459 Chr.19: 11722407-11722407 on Build GRCh38
rs755318879 Chr.19: 11722533-11722533 on Build GRCh38
rs752934112 Chr.19: 11722451-11722451 on Build GRCh38
rs144091748 Chr.19: 11722547-11722547 on Build GRCh38
rs771324985 Chr.19: 11722555-11722555 on Build GRCh38
rs369947785 Chr.19: 11722580-11722580 on Build GRCh38
rs367948038 Chr.19: 11722472-11722472 on Build GRCh38
rs757167019 Chr.19: 11722482-11722482 on Build GRCh38
rs777864795 Chr.19: 11722459-11722459 on Build GRCh38
rs760000070 Chr.19: 11722422-11722422 on Build GRCh38

View Full Product Details