Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43356082-43356335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENOX1
Entrez Gene ID
55068
Gene Name
ecto-NOX disulfide-thiol exchanger 1
Gene Aliases
CNOX, PIG38, bA64J21.1, cCNOX
UniGene
Hs.128258
Gene Chromosome Location
Chr.13: 43213525-43786980 on Build GRCh38
Transcripts
NM_001127615.1 NM_001242863.1 NM_017993.3 XM_005266439.3 XM_011535126.2 XM_011535127.2 XM_011535128.2 XM_011535132.2 XM_017020637.1 XM_017020638.1 XM_017020639.1 XM_017020640.1 XM_017020641.1 XM_017020642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274965 c.635G>A p.G212D Chr.13: 43356107-43356107 on Build GRCh38
4601459 c.593A>G p.Y198C Chr.13: 43356149-43356149 on Build GRCh38
6074636 c.607G>T p.G203W Chr.13: 43356135-43356135 on Build GRCh38
6869954 c.596G>A p.R199K Chr.13: 43356146-43356146 on Build GRCh38
432412 c.602G>A p.R201Q Chr.13: 43356140-43356140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376558847 Chr.13: 43356163-43356163 on Build GRCh38
rs763074722 Chr.13: 43356183-43356183 on Build GRCh38
rs769841758 Chr.13: 43356178-43356178 on Build GRCh38
rs761199321 Chr.13: 43356285-43356285 on Build GRCh38
rs779765900 Chr.13: 43356127-43356127 on Build GRCh38
rs753247747 Chr.13: 43356133-43356133 on Build GRCh38
rs369352426 Chr.13: 43356186-43356186 on Build GRCh38
rs543261899 Chr.13: 43356226-43356226 on Build GRCh38
rs2281713 Chr.13: 43356201-43356201 on Build GRCh38
rs1054485403 Chr.13: 43356215-43356215 on Build GRCh38
rs775809212 Chr.13: 43356179-43356179 on Build GRCh38
rs778444392 Chr.13: 43356141-43356141 on Build GRCh38
rs772062303 Chr.13: 43356159-43356159 on Build GRCh38
rs550107788 Chr.13: 43356190-43356190 on Build GRCh38
rs1037996527 Chr.13: 43356239-43356239 on Build GRCh38
rs570720118 Chr.13: 43356104-43356104 on Build GRCh38
rs1018381192 Chr.13: 43356280-43356280 on Build GRCh38
rs143923184 Chr.13: 43356124-43356124 on Build GRCh38
rs147264644 Chr.13: 43356266-43356266 on Build GRCh38
rs371824797 Chr.13: 43356106-43356106 on Build GRCh38
rs557517271 Chr.13: 43356288-43356288 on Build GRCh38
rs745499217 Chr.13: 43356310-43356310 on Build GRCh38
rs1051840204 Chr.13: 43356279-43356279 on Build GRCh38
rs746778108 Chr.13: 43356221-43356221 on Build GRCh38
rs371252384 Chr.13: 43356125-43356125 on Build GRCh38
rs192403508 Chr.13: 43356192-43356192 on Build GRCh38
rs534604067 Chr.13: 43356287-43356287 on Build GRCh38
rs754506922 Chr.13: 43356134-43356134 on Build GRCh38
rs572080955 Chr.13: 43356268-43356268 on Build GRCh38
rs777416402 Chr.13: 43356162-43356162 on Build GRCh38
rs555231135 Chr.13: 43356262-43356262 on Build GRCh38
rs183797317 Chr.13: 43356217-43356217 on Build GRCh38
rs1044818206 Chr.13: 43356149-43356149 on Build GRCh38
rs768155526 Chr.13: 43356200-43356200 on Build GRCh38
rs767059099 Chr.13: 43356265-43356265 on Build GRCh38
rs748048089 Chr.13: 43356144-43356144 on Build GRCh38
rs767992156 Chr.13: 43356118-43356118 on Build GRCh38
rs551751432 Chr.13: 43356166-43356167 on Build GRCh38

View Full Product Details