Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160123094-160123355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A2
Entrez Gene ID
477
Gene Name
ATPase Na+/K+ transporting subunit alpha 2
Gene Aliases
FHM2, MHP2
UniGene
Hs.34114
Gene Chromosome Location
Chr.1: 160115730-160143591 on Build GRCh38
Transcripts
NM_000702.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
463241 c.215G>T p.R72L Chr.1: 160123250-160123250 on Build GRCh38
3802463 c.250A>G p.T84A Chr.1: 160123285-160123285 on Build GRCh38
1580539 c.254C>A p.P85H Chr.1: 160123289-160123289 on Build GRCh38
4974980 c.280C>G p.L94V Chr.1: 160123315-160123315 on Build GRCh38
6378920 c.285C>A p.F95L Chr.1: 160123320-160123320 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763526997 Chr.1: 160123183-160123183 on Build GRCh38
rs114734823 Chr.1: 160123134-160123134 on Build GRCh38
rs1002868027 Chr.1: 160123143-160123143 on Build GRCh38
rs796052282 Chr.1: 160123227-160123227 on Build GRCh38
rs886045415 Chr.1: 160123320-160123320 on Build GRCh38
rs796052278 Chr.1: 160123309-160123309 on Build GRCh38
rs374747653 Chr.1: 160123230-160123230 on Build GRCh38
rs184873213 Chr.1: 160123186-160123186 on Build GRCh38
rs779812635 Chr.1: 160123250-160123250 on Build GRCh38
rs558323868 Chr.1: 160123240-160123240 on Build GRCh38
rs887073402 Chr.1: 160123138-160123138 on Build GRCh38
rs759727541 Chr.1: 160123189-160123189 on Build GRCh38
rs199660548 Chr.1: 160123192-160123192 on Build GRCh38
rs530467586 Chr.1: 160123133-160123133 on Build GRCh38
rs745501820 Chr.1: 160123176-160123176 on Build GRCh38
rs749280321 Chr.1: 160123170-160123170 on Build GRCh38
rs752942405 Chr.1: 160123207-160123207 on Build GRCh38
rs1057450939 Chr.1: 160123151-160123151 on Build GRCh38
rs936254331 Chr.1: 160123253-160123253 on Build GRCh38
rs764557600 Chr.1: 160123118-160123118 on Build GRCh38
rs187733403 Chr.1: 160123229-160123229 on Build GRCh38
rs777639672 Chr.1: 160123169-160123169 on Build GRCh38
rs759566498 Chr.1: 160123326-160123326 on Build GRCh38
rs755863566 Chr.1: 160123211-160123211 on Build GRCh38
rs780963435 Chr.1: 160123163-160123163 on Build GRCh38
rs1021718735 Chr.1: 160123185-160123185 on Build GRCh38
rs201540165 Chr.1: 160123263-160123263 on Build GRCh38
rs537472446 Chr.1: 160123281-160123281 on Build GRCh38
rs201356663 Chr.1: 160123177-160123177 on Build GRCh38
rs773681174 Chr.1: 160123282-160123282 on Build GRCh38
rs756027299 Chr.1: 160123167-160123167 on Build GRCh38
rs774275559 Chr.1: 160123310-160123310 on Build GRCh38
rs372105057 Chr.1: 160123206-160123206 on Build GRCh38
rs771511480 Chr.1: 160123302-160123302 on Build GRCh38
rs372138843 Chr.1: 160123131-160123131 on Build GRCh38
rs568448684 Chr.1: 160123175-160123175 on Build GRCh38
rs121918619 Chr.1: 160123228-160123228 on Build GRCh38
rs950102855 Chr.1: 160123315-160123315 on Build GRCh38
rs551196059 Chr.1: 160123128-160123128 on Build GRCh38
rs770282237 Chr.1: 160123172-160123172 on Build GRCh38
rs763362150 Chr.1: 160123283-160123283 on Build GRCh38
rs745431386 Chr.1: 160123239-160123239 on Build GRCh38
rs751731933 Chr.1: 160123188-160123188 on Build GRCh38
rs372532104 Chr.1: 160123182-160123182 on Build GRCh38

View Full Product Details