Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26320134-26320371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD52
Entrez Gene ID
1043
Gene Name
CD52 molecule
Gene Aliases
CDW52
UniGene
Hs.276770
Gene Chromosome Location
Chr.1: 26317920-26320523 on Build GRCh38
Transcripts
NM_001803.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2235092 c.158C>T p.A53V Chr.1: 26320274-26320274 on Build GRCh38
4890664 c.62C>A p.T21N Chr.1: 26320178-26320178 on Build GRCh38
3689548 c.81C>T p.N27N Chr.1: 26320197-26320197 on Build GRCh38
6623821 c.82G>A p.D28N Chr.1: 26320198-26320198 on Build GRCh38
4030949 c.79A>C p.N27H Chr.1: 26320195-26320195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776787410 Chr.1: 26320233-26320233 on Build GRCh38
rs966328320 Chr.1: 26320290-26320290 on Build GRCh38
rs763003427 Chr.1: 26320310-26320310 on Build GRCh38
rs201908321 Chr.1: 26320243-26320243 on Build GRCh38
rs763079859 Chr.1: 26320196-26320196 on Build GRCh38
rs867037545 Chr.1: 26320174-26320174 on Build GRCh38
rs766360443 Chr.1: 26320311-26320311 on Build GRCh38
rs553102200 Chr.1: 26320159-26320161 on Build GRCh38
rs1071849 Chr.1: 26320235-26320235 on Build GRCh38
rs756511544 Chr.1: 26320331-26320331 on Build GRCh38
rs761729283 Chr.1: 26320305-26320305 on Build GRCh38
rs1014021564 Chr.1: 26320157-26320157 on Build GRCh38
rs41285429 Chr.1: 26320292-26320292 on Build GRCh38
rs945648441 Chr.1: 26320303-26320303 on Build GRCh38
rs748403780 Chr.1: 26320159-26320159 on Build GRCh38
rs752270798 Chr.1: 26320328-26320328 on Build GRCh38
rs41285349 Chr.1: 26320263-26320263 on Build GRCh38
rs150540528 Chr.1: 26320190-26320190 on Build GRCh38
rs964524131 Chr.1: 26320217-26320217 on Build GRCh38
rs754442377 Chr.1: 26320325-26320325 on Build GRCh38
rs770977151 Chr.1: 26320197-26320197 on Build GRCh38
rs77417309 Chr.1: 26320192-26320192 on Build GRCh38
rs771387802 Chr.1: 26320282-26320282 on Build GRCh38
rs17645 Chr.1: 26320239-26320239 on Build GRCh38
rs575936342 Chr.1: 26320327-26320327 on Build GRCh38
rs371202375 Chr.1: 26320309-26320309 on Build GRCh38
rs950716088 Chr.1: 26320178-26320178 on Build GRCh38
rs750360671 Chr.1: 26320242-26320242 on Build GRCh38
rs765090032 Chr.1: 26320308-26320308 on Build GRCh38
rs377097614 Chr.1: 26320264-26320264 on Build GRCh38
rs749765496 Chr.1: 26320278-26320278 on Build GRCh38
rs750981479 Chr.1: 26320244-26320244 on Build GRCh38
rs567106454 Chr.1: 26320231-26320231 on Build GRCh38
rs912876350 Chr.1: 26320270-26320270 on Build GRCh38
rs996113096 Chr.1: 26320225-26320225 on Build GRCh38
rs775574862 Chr.1: 26320299-26320299 on Build GRCh38
rs774596319 Chr.1: 26320201-26320201 on Build GRCh38
rs778303094 Chr.1: 26320276-26320276 on Build GRCh38
rs375340876 Chr.1: 26320350-26320350 on Build GRCh38
rs774437535 Chr.1: 26320287-26320287 on Build GRCh38
rs1026284712 Chr.1: 26320162-26320162 on Build GRCh38
rs759479775 Chr.1: 26320291-26320291 on Build GRCh38
rs536943482 Chr.1: 26320164-26320164 on Build GRCh38
rs754672292 Chr.1: 26320245-26320245 on Build GRCh38
rs767589898 Chr.1: 26320226-26320226 on Build GRCh38
rs201785995 Chr.1: 26320212-26320212 on Build GRCh38
rs751083627 Chr.1: 26320319-26320319 on Build GRCh38
rs898677202 Chr.1: 26320207-26320207 on Build GRCh38

View Full Product Details