Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124791545-124791786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL10
Entrez Gene ID
399818
Gene Name
methyltransferase like 10
Gene Aliases
C10orf138
UniGene
Hs.720819
Gene Chromosome Location
Chr.10: 124757831-124791941 on Build GRCh38
Transcripts
NM_001304467.1 NM_001304468.1 NM_212554.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6442345 c.70G>A p.E24K Chr.10: 124791764-124791764 on Build GRCh38
6400557 c.70G>C p.E24Q Chr.10: 124791764-124791764 on Build GRCh38
5811946 c.67G>T p.G23W Chr.10: 124791767-124791767 on Build GRCh38
6365489 c.64C>A p.P22T Chr.10: 124791770-124791770 on Build GRCh38
5147333 c.110+10A>C p.? Chr.10: 124791714-124791714 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370400952 Chr.10: 124791688-124791688 on Build GRCh38
rs949044205 Chr.10: 124791642-124791642 on Build GRCh38
rs990511500 Chr.10: 124791628-124791628 on Build GRCh38
rs1008927287 Chr.10: 124791577-124791577 on Build GRCh38
rs761759176 Chr.10: 124791750-124791750 on Build GRCh38
rs569266385 Chr.10: 124791671-124791671 on Build GRCh38
rs1033248508 Chr.10: 124791751-124791751 on Build GRCh38
rs767435732 Chr.10: 124791760-124791760 on Build GRCh38
rs916362841 Chr.10: 124791636-124791636 on Build GRCh38
rs750176736 Chr.10: 124791761-124791761 on Build GRCh38
rs760896853 Chr.10: 124791731-124791731 on Build GRCh38
rs779523020 Chr.10: 124791769-124791769 on Build GRCh38
rs1004770842 Chr.10: 124791652-124791652 on Build GRCh38
rs547603463 Chr.10: 124791597-124791597 on Build GRCh38
rs1045912773 Chr.10: 124791595-124791595 on Build GRCh38
rs1037655488 Chr.10: 124791653-124791653 on Build GRCh38
rs771071018 Chr.10: 124791741-124791741 on Build GRCh38
rs565814184 Chr.10: 124791644-124791644 on Build GRCh38
rs748706491 Chr.10: 124791686-124791686 on Build GRCh38
rs536650041 Chr.10: 124791690-124791690 on Build GRCh38
rs539689968 Chr.10: 124791649-124791649 on Build GRCh38
rs751450191 Chr.10: 124791749-124791749 on Build GRCh38
rs550228554 Chr.10: 124791747-124791747 on Build GRCh38
rs74866494 Chr.10: 124791707-124791707 on Build GRCh38
rs555038065 Chr.10: 124791691-124791691 on Build GRCh38
rs377726832 Chr.10: 124791718-124791718 on Build GRCh38
rs879370454 Chr.10: 124791600-124791600 on Build GRCh38
rs940595876 Chr.10: 124791650-124791650 on Build GRCh38
rs111311843 Chr.10: 124791629-124791629 on Build GRCh38
rs1020925333 Chr.10: 124791578-124791578 on Build GRCh38
rs1001189874 Chr.10: 124791620-124791620 on Build GRCh38
rs1014737011 Chr.10: 124791657-124791657 on Build GRCh38
rs776799063 Chr.10: 124791742-124791742 on Build GRCh38
rs926118423 Chr.10: 124791617-124791617 on Build GRCh38
rs545650753 Chr.10: 124791748-124791748 on Build GRCh38
rs774670451 Chr.10: 124791719-124791725 on Build GRCh38
rs991631716 Chr.10: 124791762-124791762 on Build GRCh38
rs958903897 Chr.10: 124791618-124791618 on Build GRCh38
rs978866832 Chr.10: 124791611-124791611 on Build GRCh38
rs573445612 Chr.10: 124791695-124791695 on Build GRCh38
rs201307810 Chr.10: 124791745-124791745 on Build GRCh38
rs985416622 Chr.10: 124791719-124791719 on Build GRCh38
rs755861097 Chr.10: 124791765-124791765 on Build GRCh38
rs2277264 Chr.10: 124791664-124791664 on Build GRCh38

View Full Product Details