Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4469142-4469403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMRAL1
Entrez Gene ID
57407
Gene Name
NmrA-like family domain containing 1
Gene Aliases
HSCARG, SDR48A1
UniGene
Hs.288969
Gene Chromosome Location
Chr.16: 4461677-4476320 on Build GRCh38
Transcripts
NM_001305141.1 NM_001305142.1 NM_020677.4 XM_006720905.3 XM_006720906.2 XM_011522567.2 XM_017023485.1 XM_017023486.1 XM_017023487.1 XM_017023488.1 XM_017023489.1 XM_017023490.1 XM_017023491.1 XM_017023492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7209521 c.144G>A p.R48R Chr.16: 4469362-4469362 on Build GRCh38
6505738 c.246G>A p.W82* Chr.16: 4469260-4469260 on Build GRCh38
6727426 c.110G>A p.R37Q Chr.16: 4469396-4469396 on Build GRCh38
4655710 c.118A>T p.R40W Chr.16: 4469388-4469388 on Build GRCh38
6727429 c.205C>T p.L69L Chr.16: 4469301-4469301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771607359 Chr.16: 4469287-4469287 on Build GRCh38
rs970815075 Chr.16: 4469214-4469214 on Build GRCh38
rs28576729 Chr.16: 4469299-4469299 on Build GRCh38
rs770060966 Chr.16: 4469304-4469304 on Build GRCh38
rs765331741 Chr.16: 4469218-4469231 on Build GRCh38
rs747391973 Chr.16: 4469198-4469198 on Build GRCh38
rs376563805 Chr.16: 4469347-4469347 on Build GRCh38
rs369860675 Chr.16: 4469260-4469260 on Build GRCh38
rs781650343 Chr.16: 4469269-4469269 on Build GRCh38
rs761972139 Chr.16: 4469243-4469243 on Build GRCh38
rs760453113 Chr.16: 4469227-4469227 on Build GRCh38
rs140164470 Chr.16: 4469274-4469274 on Build GRCh38
rs778576437 Chr.16: 4469285-4469285 on Build GRCh38
rs200122362 Chr.16: 4469221-4469221 on Build GRCh38
rs1027435203 Chr.16: 4469312-4469312 on Build GRCh38
rs758491855 Chr.16: 4469249-4469249 on Build GRCh38
rs745424104 Chr.16: 4469286-4469286 on Build GRCh38
rs539716222 Chr.16: 4469233-4469233 on Build GRCh38
rs528589517 Chr.16: 4469176-4469176 on Build GRCh38
rs777944883 Chr.16: 4469379-4469379 on Build GRCh38
rs530859333 Chr.16: 4469212-4469212 on Build GRCh38
rs748182600 Chr.16: 4469296-4469296 on Build GRCh38
rs376569795 Chr.16: 4469342-4469342 on Build GRCh38
rs145571336 Chr.16: 4469196-4469196 on Build GRCh38
rs555219424 Chr.16: 4469321-4469321 on Build GRCh38
rs553535524 Chr.16: 4469238-4469238 on Build GRCh38
rs756530459 Chr.16: 4469345-4469345 on Build GRCh38
rs113380483 Chr.16: 4469364-4469364 on Build GRCh38
rs146163636 Chr.16: 4469267-4469267 on Build GRCh38
rs765276728 Chr.16: 4469178-4469178 on Build GRCh38
rs771842296 Chr.16: 4469223-4469223 on Build GRCh38
rs773418135 Chr.16: 4469310-4469310 on Build GRCh38
rs750059757 Chr.16: 4469355-4469355 on Build GRCh38
rs374640354 Chr.16: 4469335-4469335 on Build GRCh38
rs199727554 Chr.16: 4469222-4469222 on Build GRCh38
rs762987363 Chr.16: 4469318-4469318 on Build GRCh38
rs767996310 Chr.16: 4469340-4469340 on Build GRCh38
rs750446081 Chr.16: 4469248-4469248 on Build GRCh38
rs370838589 Chr.16: 4469378-4469378 on Build GRCh38
rs148847461 Chr.16: 4469381-4469381 on Build GRCh38
rs773525213 Chr.16: 4469236-4469241 on Build GRCh38
rs756900384 Chr.16: 4469275-4469275 on Build GRCh38
rs777334301 Chr.16: 4469208-4469208 on Build GRCh38
rs568531799 Chr.16: 4469204-4469204 on Build GRCh38
rs919921565 Chr.16: 4469160-4469160 on Build GRCh38
rs763527251 Chr.16: 4469372-4469372 on Build GRCh38
rs762037419 Chr.16: 4469300-4469300 on Build GRCh38
rs765307367 Chr.16: 4469244-4469244 on Build GRCh38
rs865970006 Chr.16: 4469175-4469175 on Build GRCh38
rs928683832 Chr.16: 4469354-4469354 on Build GRCh38
rs143613220 Chr.16: 4469293-4469293 on Build GRCh38
rs750962011 Chr.16: 4469187-4469187 on Build GRCh38
rs202152824 Chr.16: 4469224-4469224 on Build GRCh38
rs766463471 Chr.16: 4469320-4469320 on Build GRCh38
rs933070833 Chr.16: 4469363-4469363 on Build GRCh38
rs757893968 Chr.16: 4469361-4469361 on Build GRCh38
rs766862446 Chr.16: 4469259-4469259 on Build GRCh38
rs758766545 Chr.16: 4469195-4469195 on Build GRCh38
rs779567114 Chr.16: 4469367-4469367 on Build GRCh38
rs748929754 Chr.16: 4469210-4469210 on Build GRCh38

View Full Product Details