Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35013544-35013783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAMD1A
Entrez Gene ID
57655
Gene Name
GRAM domain containing 1A
Gene Aliases
KIAA1533
UniGene
Hs.515351
Gene Chromosome Location
Chr.19: 34994784-35026471 on Build GRCh38
Transcripts
NM_001136199.2 NM_001320034.1 NM_001320035.1 NM_001320036.1 NM_020895.4 XM_011527149.1 XM_011527153.1 XM_011527154.2 XM_011527155.1 XM_011527156.1 XM_017027034.1 XM_017027035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5741532 c.814C>T p.R272C Chr.19: 35013635-35013635 on Build GRCh38
6674758 c.748G>A p.A250T Chr.19: 35013569-35013569 on Build GRCh38
354000 c.791A>G p.Q264R Chr.19: 35013612-35013612 on Build GRCh38
4896063 c.764C>T p.T255I Chr.19: 35013585-35013585 on Build GRCh38
327441 c.868G>A p.G290R Chr.19: 35013689-35013689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039495738 Chr.19: 35013594-35013594 on Build GRCh38
rs758086727 Chr.19: 35013639-35013639 on Build GRCh38
rs760394231 Chr.19: 35013645-35013645 on Build GRCh38
rs757845825 Chr.19: 35013584-35013584 on Build GRCh38
rs748353358 Chr.19: 35013666-35013666 on Build GRCh38
rs746889317 Chr.19: 35013592-35013592 on Build GRCh38
rs200884983 Chr.19: 35013571-35013571 on Build GRCh38
rs774655097 Chr.19: 35013611-35013611 on Build GRCh38
rs767644688 Chr.19: 35013568-35013568 on Build GRCh38
rs372010823 Chr.19: 35013697-35013697 on Build GRCh38
rs777067803 Chr.19: 35013693-35013693 on Build GRCh38
rs767192306 Chr.19: 35013714-35013714 on Build GRCh38
rs1006630500 Chr.19: 35013593-35013593 on Build GRCh38
rs115996530 Chr.19: 35013676-35013676 on Build GRCh38
rs751030324 Chr.19: 35013651-35013651 on Build GRCh38
rs758393718 Chr.19: 35013591-35013592 on Build GRCh38
rs372725667 Chr.19: 35013657-35013657 on Build GRCh38
rs781114140 Chr.19: 35013653-35013653 on Build GRCh38
rs201232202 Chr.19: 35013733-35013733 on Build GRCh38
rs370589936 Chr.19: 35013631-35013631 on Build GRCh38
rs947655126 Chr.19: 35013743-35013743 on Build GRCh38
rs757268242 Chr.19: 35013595-35013595 on Build GRCh38
rs775880655 Chr.19: 35013698-35013698 on Build GRCh38
rs750877472 Chr.19: 35013569-35013569 on Build GRCh38
rs777613666 Chr.19: 35013615-35013619 on Build GRCh38
rs746800511 Chr.19: 35013709-35013710 on Build GRCh38
rs182927749 Chr.19: 35013606-35013606 on Build GRCh38
rs756156610 Chr.19: 35013726-35013726 on Build GRCh38
rs201944800 Chr.19: 35013591-35013591 on Build GRCh38
rs752353349 Chr.19: 35013583-35013583 on Build GRCh38
rs900830902 Chr.19: 35013604-35013604 on Build GRCh38
rs763248686 Chr.19: 35013699-35013699 on Build GRCh38
rs7251988 Chr.19: 35013723-35013723 on Build GRCh38
rs761326569 Chr.19: 35013710-35013710 on Build GRCh38
rs374622037 Chr.19: 35013633-35013633 on Build GRCh38
rs938797618 Chr.19: 35013680-35013680 on Build GRCh38
rs149214732 Chr.19: 35013679-35013679 on Build GRCh38
rs867628330 Chr.19: 35013588-35013588 on Build GRCh38
rs1048232910 Chr.19: 35013577-35013577 on Build GRCh38
rs755360846 Chr.19: 35013578-35013578 on Build GRCh38
rs371486274 Chr.19: 35013638-35013638 on Build GRCh38
rs760093758 Chr.19: 35013695-35013695 on Build GRCh38
rs781011646 Chr.19: 35013605-35013605 on Build GRCh38
rs754280652 Chr.19: 35013573-35013573 on Build GRCh38
rs765599980 Chr.19: 35013636-35013636 on Build GRCh38
rs371740352 Chr.19: 35013677-35013677 on Build GRCh38
rs748435155 Chr.19: 35013615-35013615 on Build GRCh38
rs770691460 Chr.19: 35013720-35013721 on Build GRCh38
rs766863713 Chr.19: 35013634-35013634 on Build GRCh38
rs971898249 Chr.19: 35013665-35013665 on Build GRCh38
rs763760136 Chr.19: 35013706-35013706 on Build GRCh38
rs201394899 Chr.19: 35013635-35013635 on Build GRCh38
rs376135948 Chr.19: 35013646-35013646 on Build GRCh38
rs779976374 Chr.19: 35013737-35013737 on Build GRCh38
rs984480246 Chr.19: 35013667-35013667 on Build GRCh38
rs755955235 Chr.19: 35013661-35013661 on Build GRCh38
rs146626930 Chr.19: 35013652-35013652 on Build GRCh38
rs772036426 Chr.19: 35013627-35013627 on Build GRCh38
rs199949479 Chr.19: 35013669-35013669 on Build GRCh38

View Full Product Details