Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143267548-143267817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTK1
Entrez Gene ID
373156
Gene Name
glutathione S-transferase kappa 1
Gene Aliases
GST, GST 13-13, GST13, GST13-13, GSTK1-1, hGSTK1
UniGene
Hs.390667
Gene Chromosome Location
Chr.7: 143263429-143269129 on Build GRCh38
Transcripts
NM_001143679.1 NM_001143680.1 NM_001143681.1 NM_015917.2 NR_133932.1

Gene Symbol
LOC105375545
Entrez Gene ID
105375545
Gene Name
uncharacterized LOC105375545
Gene Aliases
-
UniGene
Hs.699839
Gene Chromosome Location
Chr.7: 143255299-143286334 on Build GRCh38
Transcripts
NM_001143679.1 NM_001143680.1 NM_001143681.1 NM_015917.2 NR_133932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5421401 c.672C>G p.L224L Chr.7: 143267700-143267700 on Build GRCh38
3721994 c.686A>G p.E229G Chr.7: 143267714-143267714 on Build GRCh38
3257996 c.693C>T p.A231A Chr.7: 143267721-143267721 on Build GRCh38
4935987 c.634C>A p.L212M Chr.7: 143267662-143267662 on Build GRCh38
1086719 c.477C>T p.I159I Chr.7: 143267673-143267673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376062844 Chr.7: 143267754-143267754 on Build GRCh38
rs754273341 Chr.7: 143267777-143267777 on Build GRCh38
rs987658399 Chr.7: 143267785-143267785 on Build GRCh38
rs770653608 Chr.7: 143267752-143267753 on Build GRCh38
rs371499025 Chr.7: 143267686-143267686 on Build GRCh38
rs570471159 Chr.7: 143267615-143267615 on Build GRCh38
rs761457484 Chr.7: 143267730-143267730 on Build GRCh38
rs762599291 Chr.7: 143267589-143267589 on Build GRCh38
rs779857212 Chr.7: 143267685-143267685 on Build GRCh38
rs763616273 Chr.7: 143267593-143267593 on Build GRCh38
rs750893755 Chr.7: 143267643-143267643 on Build GRCh38
rs41275042 Chr.7: 143267678-143267678 on Build GRCh38
rs1053192443 Chr.7: 143267612-143267612 on Build GRCh38
rs199499365 Chr.7: 143267679-143267679 on Build GRCh38
rs941798099 Chr.7: 143267726-143267726 on Build GRCh38
rs761206789 Chr.7: 143267624-143267624 on Build GRCh38
rs973450117 Chr.7: 143267750-143267750 on Build GRCh38
rs892339580 Chr.7: 143267616-143267616 on Build GRCh38
rs746685512 Chr.7: 143267584-143267585 on Build GRCh38
rs369181149 Chr.7: 143267775-143267775 on Build GRCh38
rs756238867 Chr.7: 143267596-143267596 on Build GRCh38
rs375994498 Chr.7: 143267633-143267633 on Build GRCh38
rs754185509 Chr.7: 143267662-143267662 on Build GRCh38
rs954425285 Chr.7: 143267728-143267728 on Build GRCh38
rs934760762 Chr.7: 143267605-143267605 on Build GRCh38
rs749561195 Chr.7: 143267731-143267731 on Build GRCh38
rs748308736 Chr.7: 143267673-143267673 on Build GRCh38
rs949208708 Chr.7: 143267625-143267625 on Build GRCh38
rs905129006 Chr.7: 143267647-143267647 on Build GRCh38
rs768405417 Chr.7: 143267695-143267695 on Build GRCh38
rs773848240 Chr.7: 143267704-143267704 on Build GRCh38
rs141210391 Chr.7: 143267674-143267674 on Build GRCh38
rs756607884 Chr.7: 143267654-143267654 on Build GRCh38
rs866838052 Chr.7: 143267618-143267618 on Build GRCh38
rs761266282 Chr.7: 143267760-143267760 on Build GRCh38
rs755279975 Chr.7: 143267664-143267664 on Build GRCh38
rs113193886 Chr.7: 143267671-143267671 on Build GRCh38
rs771472201 Chr.7: 143267732-143267732 on Build GRCh38
rs775179110 Chr.7: 143267580-143267580 on Build GRCh38
rs766867252 Chr.7: 143267655-143267655 on Build GRCh38

View Full Product Details