Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102174518-102174789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1R1
Entrez Gene ID
3554
Gene Name
interleukin 1 receptor type 1
Gene Aliases
CD121A, D2S1473, IL-1R-alpha, IL1R, IL1RA, P80
UniGene
Hs.701982
Gene Chromosome Location
Chr.2: 102069638-102179874 on Build GRCh38
Transcripts
NM_000877.3 NM_001288706.1 NM_001320978.1 NM_001320980.1 NM_001320981.1 NM_001320982.1 NM_001320983.1 NM_001320984.1 NM_001320985.1 XM_005263930.4 XM_005263934.4 XM_011511115.2 XM_011511116.1 XM_011511117.1 XM_011511118.2 XM_017003989.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
714948 c.1104G>T p.R368S Chr.2: 102174699-102174699 on Build GRCh38
1690825 c.1103G>A p.R368K Chr.2: 102174698-102174698 on Build GRCh38
2980491 c.1125C>T p.L375L Chr.2: 102174720-102174720 on Build GRCh38
2980489 c.1014G>T p.M338I Chr.2: 102174609-102174609 on Build GRCh38
1690824 c.1018G>A p.G340S Chr.2: 102174613-102174613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775616233 Chr.2: 102174722-102174722 on Build GRCh38
rs766969004 Chr.2: 102174747-102174747 on Build GRCh38
rs139669898 Chr.2: 102174684-102174684 on Build GRCh38
rs35381810 Chr.2: 102174724-102174724 on Build GRCh38
rs113189759 Chr.2: 102174744-102174744 on Build GRCh38
rs771106660 Chr.2: 102174573-102174573 on Build GRCh38
rs760220490 Chr.2: 102174751-102174751 on Build GRCh38
rs777850951 Chr.2: 102174676-102174676 on Build GRCh38
rs776872699 Chr.2: 102174695-102174695 on Build GRCh38
rs761541251 Chr.2: 102174743-102174743 on Build GRCh38
rs963620710 Chr.2: 102174753-102174753 on Build GRCh38
rs769820748 Chr.2: 102174712-102174712 on Build GRCh38
rs28362304 Chr.2: 102174626-102174626 on Build GRCh38
rs759558813 Chr.2: 102174580-102174580 on Build GRCh38
rs368432710 Chr.2: 102174647-102174647 on Build GRCh38
rs775454898 Chr.2: 102174590-102174590 on Build GRCh38
rs201570254 Chr.2: 102174627-102174627 on Build GRCh38
rs767020146 Chr.2: 102174629-102174629 on Build GRCh38
rs765074051 Chr.2: 102174581-102174581 on Build GRCh38
rs3917315 Chr.2: 102174565-102174565 on Build GRCh38
rs764018672 Chr.2: 102174726-102174726 on Build GRCh38
rs200031827 Chr.2: 102174618-102174618 on Build GRCh38
rs972630123 Chr.2: 102174675-102174675 on Build GRCh38
rs199537286 Chr.2: 102174557-102174557 on Build GRCh38
rs1016828821 Chr.2: 102174729-102174729 on Build GRCh38
rs745760503 Chr.2: 102174704-102174704 on Build GRCh38
rs201455903 Chr.2: 102174554-102174554 on Build GRCh38
rs766558773 Chr.2: 102174562-102174563 on Build GRCh38
rs200588277 Chr.2: 102174549-102174549 on Build GRCh38
rs200239751 Chr.2: 102174754-102174754 on Build GRCh38
rs770854818 Chr.2: 102174690-102174690 on Build GRCh38
rs773809282 Chr.2: 102174735-102174735 on Build GRCh38
rs201085481 Chr.2: 102174642-102174642 on Build GRCh38
rs747044655 Chr.2: 102174562-102174562 on Build GRCh38
rs755505492 Chr.2: 102174643-102174643 on Build GRCh38
rs748323960 Chr.2: 102174552-102174552 on Build GRCh38
rs758852011 Chr.2: 102174674-102174674 on Build GRCh38
rs560745476 Chr.2: 102174737-102174737 on Build GRCh38
rs753714802 Chr.2: 102174572-102174574 on Build GRCh38
rs773525911 Chr.2: 102174561-102174561 on Build GRCh38
rs201805817 Chr.2: 102174607-102174607 on Build GRCh38
rs200109386 Chr.2: 102174637-102174637 on Build GRCh38
rs148175475 Chr.2: 102174614-102174614 on Build GRCh38
rs534082578 Chr.2: 102174564-102174565 on Build GRCh38
rs749971173 Chr.2: 102174749-102174749 on Build GRCh38
rs200984882 Chr.2: 102174574-102174574 on Build GRCh38
rs751286025 Chr.2: 102174624-102174624 on Build GRCh38
rs886664188 Chr.2: 102174692-102174692 on Build GRCh38
rs748432829 Chr.2: 102174652-102174652 on Build GRCh38

View Full Product Details