Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37568178-37568449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNL2
Entrez Gene ID
29889
Gene Name
G protein nucleolar 2
Gene Aliases
HUMAUANTIG, NGP1, Ngp-1, Nog2, Nug2
UniGene
Hs.75528
Gene Chromosome Location
Chr.1: 37566812-37595985 on Build GRCh38
Transcripts
NM_001323623.1 NM_001323624.1 NM_013285.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5938660 c.1907C>T p.P636L Chr.1: 37568319-37568319 on Build GRCh38
281420 c.1920A>G p.R640R Chr.1: 37568306-37568306 on Build GRCh38
4532421 c.1880G>A p.G627E Chr.1: 37568346-37568346 on Build GRCh38
6507197 c.1878G>T p.K626N Chr.1: 37568348-37568348 on Build GRCh38
3785280 c.1875C>T p.S625S Chr.1: 37568351-37568351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929453334 Chr.1: 37568272-37568272 on Build GRCh38
rs148472157 Chr.1: 37568329-37568329 on Build GRCh38
rs777892437 Chr.1: 37568353-37568353 on Build GRCh38
rs758262856 Chr.1: 37568334-37568334 on Build GRCh38
rs887824216 Chr.1: 37568309-37568309 on Build GRCh38
rs774754815 Chr.1: 37568265-37568265 on Build GRCh38
rs1055509441 Chr.1: 37568370-37568370 on Build GRCh38
rs200368045 Chr.1: 37568371-37568371 on Build GRCh38
rs919465646 Chr.1: 37568267-37568267 on Build GRCh38
rs769034959 Chr.1: 37568400-37568400 on Build GRCh38
rs761117413 Chr.1: 37568283-37568283 on Build GRCh38
rs748370987 Chr.1: 37568235-37568235 on Build GRCh38
rs376485294 Chr.1: 37568247-37568247 on Build GRCh38
rs866515121 Chr.1: 37568332-37568332 on Build GRCh38
rs553019517 Chr.1: 37568396-37568396 on Build GRCh38
rs934341910 Chr.1: 37568209-37568209 on Build GRCh38
rs750372408 Chr.1: 37568333-37568333 on Build GRCh38
rs998610979 Chr.1: 37568413-37568413 on Build GRCh38
rs776169632 Chr.1: 37568401-37568401 on Build GRCh38
rs536460647 Chr.1: 37568393-37568393 on Build GRCh38
rs764754494 Chr.1: 37568331-37568331 on Build GRCh38
rs749489903 Chr.1: 37568361-37568361 on Build GRCh38
rs779081167 Chr.1: 37568392-37568392 on Build GRCh38
rs377097691 Chr.1: 37568397-37568397 on Build GRCh38
rs757511990 Chr.1: 37568240-37568240 on Build GRCh38
rs753662998 Chr.1: 37568251-37568252 on Build GRCh38
rs769973463 Chr.1: 37568237-37568237 on Build GRCh38
rs751410365 Chr.1: 37568342-37568342 on Build GRCh38
rs772581299 Chr.1: 37568279-37568279 on Build GRCh38
rs77187843 Chr.1: 37568398-37568398 on Build GRCh38
rs370049441 Chr.1: 37568306-37568306 on Build GRCh38
rs193229601 Chr.1: 37568210-37568210 on Build GRCh38
rs775932893 Chr.1: 37568282-37568282 on Build GRCh38
rs373710378 Chr.1: 37568352-37568352 on Build GRCh38
rs746331686 Chr.1: 37568274-37568274 on Build GRCh38
rs747916550 Chr.1: 37568216-37568216 on Build GRCh38
rs924169643 Chr.1: 37568207-37568207 on Build GRCh38
rs766296401 Chr.1: 37568335-37568335 on Build GRCh38
rs763712619 Chr.1: 37568302-37568302 on Build GRCh38
rs749371927 Chr.1: 37568243-37568243 on Build GRCh38

View Full Product Details