Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 46134024-46134297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCP1
Entrez Gene ID
3936
Gene Name
lymphocyte cytosolic protein 1
Gene Aliases
CP64, HEL-S-37, L-PLASTIN, LC64P, LPL, PLS2
UniGene
Hs.381099
Gene Chromosome Location
Chr.13: 46125923-46182324 on Build GRCh38
Transcripts
NM_002298.4 XM_005266374.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6456083 c.1521C>T p.L507L Chr.13: 46134232-46134232 on Build GRCh38
696506 c.1539C>T p.G513G Chr.13: 46134214-46134214 on Build GRCh38
3469098 c.1518C>T p.I506I Chr.13: 46134235-46134235 on Build GRCh38
3469097 c.1626G>A p.K542K Chr.13: 46134127-46134127 on Build GRCh38
1300218 c.1575G>A p.V525V Chr.13: 46134178-46134178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751587269 Chr.13: 46134195-46134195 on Build GRCh38
rs771620628 Chr.13: 46134254-46134254 on Build GRCh38
rs749367534 Chr.13: 46134214-46134214 on Build GRCh38
rs74073559 Chr.13: 46134063-46134063 on Build GRCh38
rs762012540 Chr.13: 46134196-46134196 on Build GRCh38
rs753417420 Chr.13: 46134223-46134223 on Build GRCh38
rs778541280 Chr.13: 46134131-46134131 on Build GRCh38
rs780533886 Chr.13: 46134219-46134219 on Build GRCh38
rs910320651 Chr.13: 46134201-46134201 on Build GRCh38
rs768084208 Chr.13: 46134203-46134203 on Build GRCh38
rs764238982 Chr.13: 46134194-46134194 on Build GRCh38
rs781658701 Chr.13: 46134117-46134117 on Build GRCh38
rs763296994 Chr.13: 46134180-46134180 on Build GRCh38
rs778462931 Chr.13: 46134238-46134238 on Build GRCh38
rs111548644 Chr.13: 46134178-46134178 on Build GRCh38
rs756565294 Chr.13: 46134105-46134105 on Build GRCh38
rs145656700 Chr.13: 46134152-46134152 on Build GRCh38
rs748338956 Chr.13: 46134144-46134144 on Build GRCh38
rs4941543 Chr.13: 46134156-46134156 on Build GRCh38
rs779826960 Chr.13: 46134107-46134107 on Build GRCh38
rs750775385 Chr.13: 46134101-46134101 on Build GRCh38
rs148900496 Chr.13: 46134141-46134141 on Build GRCh38
rs982124082 Chr.13: 46134119-46134119 on Build GRCh38
rs962551899 Chr.13: 46134228-46134228 on Build GRCh38
rs754599067 Chr.13: 46134232-46134232 on Build GRCh38
rs747610301 Chr.13: 46134253-46134253 on Build GRCh38
rs9595416 Chr.13: 46134095-46134095 on Build GRCh38
rs773155889 Chr.13: 46134145-46134145 on Build GRCh38
rs541668480 Chr.13: 46134081-46134081 on Build GRCh38
rs781135662 Chr.13: 46134109-46134109 on Build GRCh38
rs530085015 Chr.13: 46134122-46134122 on Build GRCh38
rs781764871 Chr.13: 46134088-46134088 on Build GRCh38
rs148476191 Chr.13: 46134113-46134113 on Build GRCh38
rs867594237 Chr.13: 46134231-46134231 on Build GRCh38
rs750132542 Chr.13: 46134104-46134104 on Build GRCh38
rs751013494 Chr.13: 46134205-46134205 on Build GRCh38

View Full Product Details