Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 143886612-143886873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC2HC1B
Entrez Gene ID
153918
Gene Name
zinc finger C2HC-type containing 1B
Gene Aliases
C6orf94, FAM164B, dJ468K18.5
UniGene
Hs.121188
Gene Chromosome Location
Chr.6: 143864436-143938346 on Build GRCh38
Transcripts
NM_001013623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6012523 c.42C>T p.P14P Chr.6: 143886802-143886802 on Build GRCh38
4488636 c.47C>T p.P16L Chr.6: 143886807-143886807 on Build GRCh38
3017138 c.43C>A p.P15T Chr.6: 143886803-143886803 on Build GRCh38
5404503 c.28C>T p.L10F Chr.6: 143886788-143886788 on Build GRCh38
5031471 c.35C>A p.P12H Chr.6: 143886795-143886795 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs978158278 Chr.6: 143886749-143886749 on Build GRCh38
rs985574683 Chr.6: 143886702-143886702 on Build GRCh38
rs979182905 Chr.6: 143886842-143886842 on Build GRCh38
rs779455956 Chr.6: 143886785-143886785 on Build GRCh38
rs995181563 Chr.6: 143886679-143886680 on Build GRCh38
rs961154897 Chr.6: 143886644-143886644 on Build GRCh38
rs780500695 Chr.6: 143886833-143886833 on Build GRCh38
rs377068265 Chr.6: 143886680-143886680 on Build GRCh38
rs967366050 Chr.6: 143886795-143886795 on Build GRCh38
rs899096270 Chr.6: 143886686-143886686 on Build GRCh38
rs1001357736 Chr.6: 143886761-143886761 on Build GRCh38
rs936747172 Chr.6: 143886780-143886780 on Build GRCh38
rs1034210380 Chr.6: 143886770-143886770 on Build GRCh38
rs751268266 Chr.6: 143886721-143886721 on Build GRCh38
rs370012101 Chr.6: 143886743-143886743 on Build GRCh38
rs770924167 Chr.6: 143886739-143886739 on Build GRCh38
rs868033051 Chr.6: 143886692-143886692 on Build GRCh38
rs966004980 Chr.6: 143886744-143886744 on Build GRCh38
rs755615708 Chr.6: 143886783-143886783 on Build GRCh38
rs762702431 Chr.6: 143886679-143886679 on Build GRCh38
rs1014374936 Chr.6: 143886782-143886782 on Build GRCh38
rs752206232 Chr.6: 143886764-143886764 on Build GRCh38
rs767098071 Chr.6: 143886763-143886763 on Build GRCh38
rs768727403 Chr.6: 143886794-143886794 on Build GRCh38
rs1053842875 Chr.6: 143886801-143886801 on Build GRCh38
rs575686777 Chr.6: 143886754-143886754 on Build GRCh38
rs746365328 Chr.6: 143886806-143886806 on Build GRCh38
rs758729952 Chr.6: 143886831-143886831 on Build GRCh38
rs867067208 Chr.6: 143886773-143886773 on Build GRCh38

View Full Product Details