Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36638929-36639154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF461
Entrez Gene ID
92283
Gene Name
zinc finger protein 461
Gene Aliases
GIOT-1, GIOT1, HZF28
UniGene
Hs.590972
Gene Chromosome Location
Chr.19: 36636621-36666853 on Build GRCh38
Transcripts
NM_001297623.2 NM_001322821.1 NM_001322823.1 NM_001322825.1 NM_001322826.1 NM_001322827.1 NM_001322828.1 NM_153257.4 XM_011527486.2 XM_017027470.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6465725 c.1248T>G p.H416Q Chr.19: 36639097-36639097 on Build GRCh38
4752942 c.1035G>A p.M345I Chr.19: 36638929-36638929 on Build GRCh38
6150350 c.997C>T p.Q333* Chr.19: 36638967-36638967 on Build GRCh38
995526 c.1301T>G p.I434S Chr.19: 36639044-36639044 on Build GRCh38
4077418 c.1211A>G p.Q404R Chr.19: 36639134-36639134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182036940 Chr.19: 36639116-36639116 on Build GRCh38
rs776334362 Chr.19: 36639087-36639093 on Build GRCh38
rs764102765 Chr.19: 36638984-36638984 on Build GRCh38
rs545135367 Chr.19: 36639106-36639106 on Build GRCh38
rs557545377 Chr.19: 36638952-36638952 on Build GRCh38
rs573976946 Chr.19: 36638955-36638955 on Build GRCh38
rs772160829 Chr.19: 36639045-36639045 on Build GRCh38
rs368670478 Chr.19: 36639055-36639055 on Build GRCh38
rs759744718 Chr.19: 36638954-36638954 on Build GRCh38
rs969483127 Chr.19: 36639131-36639131 on Build GRCh38
rs776693702 Chr.19: 36638953-36638953 on Build GRCh38
rs559189273 Chr.19: 36639065-36639065 on Build GRCh38
rs544554907 Chr.19: 36638987-36638987 on Build GRCh38
rs747446633 Chr.19: 36639077-36639077 on Build GRCh38
rs755746967 Chr.19: 36639038-36639038 on Build GRCh38
rs990083887 Chr.19: 36639073-36639073 on Build GRCh38
rs762696368 Chr.19: 36638975-36638975 on Build GRCh38
rs752496308 Chr.19: 36638966-36638966 on Build GRCh38
rs778245783 Chr.19: 36639071-36639071 on Build GRCh38
rs780785819 Chr.19: 36638999-36638999 on Build GRCh38
rs533176383 Chr.19: 36639017-36639017 on Build GRCh38
rs370979336 Chr.19: 36639089-36639089 on Build GRCh38
rs758744142 Chr.19: 36639058-36639058 on Build GRCh38
rs770027313 Chr.19: 36639125-36639125 on Build GRCh38
rs756824933 Chr.19: 36638993-36638993 on Build GRCh38
rs562652882 Chr.19: 36639007-36639007 on Build GRCh38

View Full Product Details