Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 135208799-135209054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZRANB3
Entrez Gene ID
84083
Gene Name
zinc finger RANBP2-type containing 3
Gene Aliases
4933425L19Rik, AH2
UniGene
Hs.658422
Gene Chromosome Location
Chr.2: 135164703-135531322 on Build GRCh38
Transcripts
NM_001286568.1 NM_001286569.1 NM_032143.3 XM_005263809.1 XM_006712788.1 XM_011511958.2 XM_011511959.2 XM_011511960.1 XM_011511961.2 XM_011511963.1 XM_011511966.2 XM_017005065.1 XM_017005066.1 XM_017005067.1 XM_017005068.1 XM_017005069.1 XM_017005070.1 XM_017005071.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4745374 c.980G>A p.R327Q Chr.2: 135208883-135208883 on Build GRCh38
4745377 c.945C>G p.G315G Chr.2: 135208918-135208918 on Build GRCh38
4814271 c.2490-1G>T p.? Chr.2: 135208979-135208979 on Build GRCh38
5937070 c.994A>C p.K332Q Chr.2: 135208869-135208869 on Build GRCh38
475929 c.904G>C p.V302L Chr.2: 135208959-135208959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377722888 Chr.2: 135208910-135208910 on Build GRCh38
rs779488550 Chr.2: 135209005-135209005 on Build GRCh38
rs762128087 Chr.2: 135208883-135208883 on Build GRCh38
rs929481762 Chr.2: 135208896-135208896 on Build GRCh38
rs564039843 Chr.2: 135208937-135208937 on Build GRCh38
rs753718080 Chr.2: 135208884-135208884 on Build GRCh38
rs1047850328 Chr.2: 135208900-135208900 on Build GRCh38
rs997510669 Chr.2: 135208966-135208966 on Build GRCh38
rs768316771 Chr.2: 135208921-135208921 on Build GRCh38
rs758805409 Chr.2: 135208981-135208981 on Build GRCh38
rs779725032 Chr.2: 135208919-135208919 on Build GRCh38
rs955236293 Chr.2: 135208978-135208978 on Build GRCh38
rs767857412 Chr.2: 135208960-135208960 on Build GRCh38
rs376147360 Chr.2: 135209010-135209010 on Build GRCh38
rs1011730408 Chr.2: 135208879-135208879 on Build GRCh38
rs751873684 Chr.2: 135208851-135208851 on Build GRCh38
rs757902241 Chr.2: 135208993-135208993 on Build GRCh38
rs770967363 Chr.2: 135208948-135208948 on Build GRCh38
rs763060159 Chr.2: 135208945-135208945 on Build GRCh38
rs773272710 Chr.2: 135208842-135208842 on Build GRCh38
rs566144005 Chr.2: 135208968-135208968 on Build GRCh38
rs547624935 Chr.2: 135208954-135208954 on Build GRCh38
rs900527075 Chr.2: 135208959-135208959 on Build GRCh38
rs749935825 Chr.2: 135208915-135208915 on Build GRCh38
rs1046001054 Chr.2: 135208832-135208832 on Build GRCh38
rs186820943 Chr.2: 135208944-135208944 on Build GRCh38
rs1001789323 Chr.2: 135208916-135208916 on Build GRCh38
rs367547440 Chr.2: 135209020-135209020 on Build GRCh38
rs746465834 Chr.2: 135208981-135208981 on Build GRCh38
rs757912143 Chr.2: 135208918-135208918 on Build GRCh38
rs750365952 Chr.2: 135208991-135208991 on Build GRCh38
rs781492529 Chr.2: 135208923-135208923 on Build GRCh38
rs757151946 Chr.2: 135208899-135208899 on Build GRCh38
rs753629147 Chr.2: 135208964-135208964 on Build GRCh38
rs779545700 Chr.2: 135208919-135208919 on Build GRCh38
rs765216225 Chr.2: 135208990-135208990 on Build GRCh38
rs770273015 Chr.2: 135208925-135208925 on Build GRCh38
rs774597691 Chr.2: 135208953-135208953 on Build GRCh38
rs973157372 Chr.2: 135208988-135208988 on Build GRCh38
rs371392862 Chr.2: 135208924-135208924 on Build GRCh38
rs754554444 Chr.2: 135209026-135209026 on Build GRCh38
rs1029594155 Chr.2: 135208970-135208970 on Build GRCh38
rs768143057 Chr.2: 135209021-135209021 on Build GRCh38
rs763005671 Chr.2: 135208843-135208843 on Build GRCh38
rs757948735 Chr.2: 135208843-135208847 on Build GRCh38
rs760526135 Chr.2: 135208863-135208863 on Build GRCh38
rs766622783 Chr.2: 135208844-135208844 on Build GRCh38
rs373709261 Chr.2: 135208836-135208836 on Build GRCh38

View Full Product Details