Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 72358092-72358319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTCD2
Entrez Gene ID
79810
Gene Name
pentatricopeptide repeat domain 2
Gene Aliases
-
UniGene
Hs.126906
Gene Chromosome Location
Chr.5: 72320367-72368395 on Build GRCh38
Transcripts
NM_001284403.1 NM_001284404.1 NM_001284405.1 NM_024754.4 XM_005248601.2 XM_005248603.2 XM_017009866.1 XM_017009867.1 XM_017009868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3429613 c.995T>G p.F332C Chr.5: 72358255-72358255 on Build GRCh38
6238621 c.962A>G p.K321R Chr.5: 72358222-72358222 on Build GRCh38
5014499 c.979G>T p.A327S Chr.5: 72358239-72358239 on Build GRCh38
267282 c.980C>T p.A327V Chr.5: 72358240-72358240 on Build GRCh38
1438462 c.997G>T p.D333Y Chr.5: 72358257-72358257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956054765 Chr.5: 72358192-72358192 on Build GRCh38
rs756818887 Chr.5: 72358179-72358179 on Build GRCh38
rs200810438 Chr.5: 72358280-72358280 on Build GRCh38
rs747157347 Chr.5: 72358276-72358276 on Build GRCh38
rs560172136 Chr.5: 72358230-72358230 on Build GRCh38
rs751168543 Chr.5: 72358161-72358161 on Build GRCh38
rs374801120 Chr.5: 72358233-72358233 on Build GRCh38
rs112960229 Chr.5: 72358236-72358236 on Build GRCh38
rs755537877 Chr.5: 72358241-72358241 on Build GRCh38
rs779389045 Chr.5: 72358247-72358247 on Build GRCh38
rs376088173 Chr.5: 72358259-72358259 on Build GRCh38
rs749643837 Chr.5: 72358234-72358234 on Build GRCh38
rs762835626 Chr.5: 72358153-72358153 on Build GRCh38
rs199700973 Chr.5: 72358192-72358193 on Build GRCh38
rs764098783 Chr.5: 72358155-72358155 on Build GRCh38
rs370784805 Chr.5: 72358221-72358221 on Build GRCh38
rs767294760 Chr.5: 72358210-72358210 on Build GRCh38
rs1019587939 Chr.5: 72358143-72358143 on Build GRCh38
rs748229887 Chr.5: 72358248-72358248 on Build GRCh38
rs770664614 Chr.5: 72358156-72358156 on Build GRCh38
rs112670257 Chr.5: 72358263-72358263 on Build GRCh38

View Full Product Details