Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 69480697-69480946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UGT2B4
Entrez Gene ID
7363
Gene Name
UDP glucuronosyltransferase family 2 member B4
Gene Aliases
HLUG25, UDPGT2B4, UDPGTH1, UDPGTh-1, UGT2B11
UniGene
Hs.285887
Gene Chromosome Location
Chr.4: 69480165-69526014 on Build GRCh38
Transcripts
NM_001297615.1 NM_001297616.1 NM_021139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1430699 c.1406G>A p.R469H Chr.4: 69480815-69480815 on Build GRCh38
6264452 c.1325C>T p.A442V Chr.4: 69480896-69480896 on Build GRCh38
1430698 c.1444G>A p.D482N Chr.4: 69480777-69480777 on Build GRCh38
286285 c.1378G>A p.A460T Chr.4: 69480843-69480843 on Build GRCh38
316353 c.1456T>G p.F486V Chr.4: 69480765-69480765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs13119049 Chr.4: 69480847-69480847 on Build GRCh38
rs375220784 Chr.4: 69480893-69480893 on Build GRCh38
rs527905109 Chr.4: 69480739-69480739 on Build GRCh38
rs67904882 Chr.4: 69480846-69480847 on Build GRCh38
rs751139318 Chr.4: 69480882-69480882 on Build GRCh38
rs371915543 Chr.4: 69480916-69480916 on Build GRCh38
rs762114586 Chr.4: 69480776-69480776 on Build GRCh38
rs758118729 Chr.4: 69480740-69480740 on Build GRCh38
rs550152016 Chr.4: 69480836-69480836 on Build GRCh38
rs201984562 Chr.4: 69480898-69480898 on Build GRCh38
rs769787957 Chr.4: 69480897-69480897 on Build GRCh38
rs772384251 Chr.4: 69480753-69480753 on Build GRCh38
rs775856632 Chr.4: 69480875-69480875 on Build GRCh38
rs184647368 Chr.4: 69480863-69480863 on Build GRCh38
rs200280202 Chr.4: 69480762-69480762 on Build GRCh38
rs764727376 Chr.4: 69480857-69480857 on Build GRCh38
rs770843089 Chr.4: 69480747-69480747 on Build GRCh38
rs368457483 Chr.4: 69480903-69480903 on Build GRCh38
rs71599906 Chr.4: 69480911-69480911 on Build GRCh38
rs367620350 Chr.4: 69480841-69480841 on Build GRCh38
rs764240029 Chr.4: 69480764-69480764 on Build GRCh38
rs767837637 Chr.4: 69480777-69480777 on Build GRCh38
rs745868845 Chr.4: 69480895-69480895 on Build GRCh38
rs1043998951 Chr.4: 69480814-69480814 on Build GRCh38
rs200320318 Chr.4: 69480816-69480816 on Build GRCh38
rs901641166 Chr.4: 69480749-69480750 on Build GRCh38
rs745780826 Chr.4: 69480835-69480835 on Build GRCh38
rs1028890192 Chr.4: 69480782-69480782 on Build GRCh38
rs747685299 Chr.4: 69480909-69480909 on Build GRCh38
rs770803752 Chr.4: 69480829-69480829 on Build GRCh38
rs980930084 Chr.4: 69480886-69480886 on Build GRCh38
rs868043600 Chr.4: 69480884-69480884 on Build GRCh38
rs957075745 Chr.4: 69480894-69480894 on Build GRCh38
rs951964641 Chr.4: 69480791-69480791 on Build GRCh38
rs774765310 Chr.4: 69480775-69480775 on Build GRCh38
rs368535155 Chr.4: 69480827-69480827 on Build GRCh38
rs564297645 Chr.4: 69480874-69480874 on Build GRCh38
rs879463290 Chr.4: 69480846-69480847 on Build GRCh38
rs778378649 Chr.4: 69480792-69480792 on Build GRCh38
rs760028353 Chr.4: 69480850-69480850 on Build GRCh38
rs547579888 Chr.4: 69480778-69480778 on Build GRCh38
rs772218125 Chr.4: 69480907-69480907 on Build GRCh38
rs747860734 Chr.4: 69480807-69480807 on Build GRCh38
rs756596732 Chr.4: 69480779-69480779 on Build GRCh38
rs567459900 Chr.4: 69480787-69480787 on Build GRCh38
rs773055119 Chr.4: 69480729-69480729 on Build GRCh38
rs374388378 Chr.4: 69480840-69480840 on Build GRCh38
rs1005885167 Chr.4: 69480798-69480798 on Build GRCh38
rs757127114 Chr.4: 69480885-69480885 on Build GRCh38
rs376320686 Chr.4: 69480812-69480812 on Build GRCh38
rs13142440 Chr.4: 69480846-69480846 on Build GRCh38
rs558607262 Chr.4: 69480870-69480870 on Build GRCh38
rs776668037 Chr.4: 69480832-69480832 on Build GRCh38
rs766863857 Chr.4: 69480781-69480781 on Build GRCh38

View Full Product Details