Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34634920-34635151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM132E
Entrez Gene ID
124842
Gene Name
transmembrane protein 132E
Gene Aliases
DFNB99
UniGene
Hs.310482
Gene Chromosome Location
Chr.17: 34580749-34639318 on Build GRCh38
Transcripts
NM_001304438.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6080604 c.1590G>A p.W530* Chr.17: 34634970-34634970 on Build GRCh38
5689124 c.1701C>G p.P567P Chr.17: 34635081-34635081 on Build GRCh38
6080605 c.1569C>T p.V523V Chr.17: 34634949-34634949 on Build GRCh38
6080603 c.1671G>T p.T557T Chr.17: 34635051-34635051 on Build GRCh38
137099 c.1552G>A p.G518S Chr.17: 34634932-34634932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907920171 Chr.17: 34634967-34634967 on Build GRCh38
rs572515209 Chr.17: 34634938-34634938 on Build GRCh38
rs111689642 Chr.17: 34635045-34635045 on Build GRCh38
rs376500448 Chr.17: 34635016-34635016 on Build GRCh38
rs752049579 Chr.17: 34635023-34635023 on Build GRCh38
rs1010124250 Chr.17: 34635061-34635061 on Build GRCh38
rs764835141 Chr.17: 34635014-34635014 on Build GRCh38
rs1038374590 Chr.17: 34635009-34635009 on Build GRCh38
rs769934251 Chr.17: 34634975-34634975 on Build GRCh38
rs530311078 Chr.17: 34635112-34635112 on Build GRCh38
rs757359081 Chr.17: 34634941-34634941 on Build GRCh38
rs374567511 Chr.17: 34635129-34635129 on Build GRCh38
rs146893987 Chr.17: 34635051-34635051 on Build GRCh38
rs778747922 Chr.17: 34634978-34634978 on Build GRCh38
rs770384316 Chr.17: 34635007-34635007 on Build GRCh38
rs150291867 Chr.17: 34635028-34635028 on Build GRCh38
rs370949147 Chr.17: 34635126-34635126 on Build GRCh38
rs376446244 Chr.17: 34634973-34634973 on Build GRCh38
rs547154694 Chr.17: 34635127-34635127 on Build GRCh38
rs200998663 Chr.17: 34635008-34635008 on Build GRCh38
rs73988752 Chr.17: 34634985-34634985 on Build GRCh38
rs776173332 Chr.17: 34635066-34635066 on Build GRCh38
rs1008020743 Chr.17: 34635047-34635047 on Build GRCh38
rs774950867 Chr.17: 34635097-34635097 on Build GRCh38
rs143181955 Chr.17: 34635080-34635080 on Build GRCh38
rs747958553 Chr.17: 34634981-34634981 on Build GRCh38
rs746896935 Chr.17: 34635057-34635057 on Build GRCh38
rs774878102 Chr.17: 34635119-34635119 on Build GRCh38
rs759201676 Chr.17: 34635011-34635011 on Build GRCh38
rs764571058 Chr.17: 34635022-34635022 on Build GRCh38
rs139247454 Chr.17: 34635063-34635063 on Build GRCh38
rs561353645 Chr.17: 34635094-34635094 on Build GRCh38
rs764517750 Chr.17: 34635117-34635117 on Build GRCh38
rs140125786 Chr.17: 34635015-34635015 on Build GRCh38
rs996407997 Chr.17: 34635124-34635124 on Build GRCh38
rs914467657 Chr.17: 34634983-34634983 on Build GRCh38
rs780197772 Chr.17: 34635039-34635039 on Build GRCh38
rs758121847 Chr.17: 34635050-34635050 on Build GRCh38
rs781427706 Chr.17: 34634953-34634953 on Build GRCh38
rs764749321 Chr.17: 34634975-34635007 on Build GRCh38
rs75594429 Chr.17: 34635031-34635031 on Build GRCh38
rs760610543 Chr.17: 34635001-34635001 on Build GRCh38
rs750520343 Chr.17: 34635029-34635029 on Build GRCh38
rs757870856 Chr.17: 34635027-34635027 on Build GRCh38
rs773075998 Chr.17: 34634986-34634986 on Build GRCh38
rs751678184 Chr.17: 34634939-34634939 on Build GRCh38

View Full Product Details