Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57720515-57720780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF671
Entrez Gene ID
79891
Gene Name
zinc finger protein 671
Gene Aliases
-
UniGene
Hs.180402
Gene Chromosome Location
Chr.19: 57719747-57727637 on Build GRCh38
Transcripts
NM_001321375.1 NM_001321376.1 NM_024833.2 XM_017027314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6152567 c.1553G>A p.R518Q Chr.19: 57720533-57720533 on Build GRCh38
3673038 c.1537G>T p.G513W Chr.19: 57720549-57720549 on Build GRCh38
1002447 c.1478A>G p.N493S Chr.19: 57720608-57720608 on Build GRCh38
2973007 c.1435T>C p.Y479H Chr.19: 57720651-57720651 on Build GRCh38
4744629 c.1432C>T p.P478S Chr.19: 57720654-57720654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs207477287 Chr.19: 57720550-57720550 on Build GRCh38
rs1027427866 Chr.19: 57720739-57720739 on Build GRCh38
rs754494653 Chr.19: 57720743-57720743 on Build GRCh38
rs751429353 Chr.19: 57720565-57720565 on Build GRCh38
rs779588583 Chr.19: 57720596-57720596 on Build GRCh38
rs866244505 Chr.19: 57720656-57720656 on Build GRCh38
rs750357486 Chr.19: 57720581-57720581 on Build GRCh38
rs776921926 Chr.19: 57720663-57720663 on Build GRCh38
rs377494586 Chr.19: 57720727-57720727 on Build GRCh38
rs200134520 Chr.19: 57720633-57720633 on Build GRCh38
rs946581603 Chr.19: 57720554-57720554 on Build GRCh38
rs201753059 Chr.19: 57720634-57720634 on Build GRCh38
rs201295756 Chr.19: 57720598-57720598 on Build GRCh38
rs771536597 Chr.19: 57720652-57720652 on Build GRCh38
rs866820452 Chr.19: 57720674-57720674 on Build GRCh38
rs767236395 Chr.19: 57720697-57720697 on Build GRCh38
rs1003171436 Chr.19: 57720591-57720591 on Build GRCh38
rs777385377 Chr.19: 57720717-57720718 on Build GRCh38
rs766592424 Chr.19: 57720552-57720552 on Build GRCh38
rs748765310 Chr.19: 57720597-57720597 on Build GRCh38
rs765351697 Chr.19: 57720690-57720690 on Build GRCh38
rs756085117 Chr.19: 57720722-57720722 on Build GRCh38
rs775580998 Chr.19: 57720692-57720692 on Build GRCh38
rs757017541 Chr.19: 57720569-57720569 on Build GRCh38
rs763196480 Chr.19: 57720694-57720694 on Build GRCh38
rs765552638 Chr.19: 57720561-57720561 on Build GRCh38
rs897526765 Chr.19: 57720684-57720684 on Build GRCh38
rs140341109 Chr.19: 57720549-57720549 on Build GRCh38
rs755262339 Chr.19: 57720656-57720656 on Build GRCh38
rs753949668 Chr.19: 57720553-57720553 on Build GRCh38
rs144165618 Chr.19: 57720576-57720576 on Build GRCh38
rs778519925 Chr.19: 57720620-57720620 on Build GRCh38
rs201258792 Chr.19: 57720661-57720661 on Build GRCh38
rs751949930 Chr.19: 57720721-57720721 on Build GRCh38
rs762075516 Chr.19: 57720543-57720543 on Build GRCh38
rs200410098 Chr.19: 57720719-57720719 on Build GRCh38
rs887930410 Chr.19: 57720745-57720745 on Build GRCh38
rs773579017 Chr.19: 57720649-57720649 on Build GRCh38
rs759882061 Chr.19: 57720556-57720556 on Build GRCh38
rs571634975 Chr.19: 57720547-57720547 on Build GRCh38
rs1056541017 Chr.19: 57720630-57720630 on Build GRCh38
rs753400550 Chr.19: 57720734-57720734 on Build GRCh38
rs573129538 Chr.19: 57720679-57720679 on Build GRCh38
rs746783414 Chr.19: 57720733-57720734 on Build GRCh38

View Full Product Details