Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35825687-35825934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGO4
Entrez Gene ID
192670
Gene Name
argonaute 4, RISC catalytic component
Gene Aliases
EIF2C4
UniGene
Hs.744858
Gene Chromosome Location
Chr.1: 35807630-35857890 on Build GRCh38
Transcripts
NM_017629.3 XM_005270578.3 XM_005270579.3 XM_011540883.2 XM_011540884.2 XM_017000529.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1687454 c.523C>T p.P175S Chr.1: 35825713-35825713 on Build GRCh38
7354805 c.526G>C p.E176Q Chr.1: 35825716-35825716 on Build GRCh38
1251074 c.506G>A p.R169H Chr.1: 35825696-35825696 on Build GRCh38
5769904 c.559G>T p.E187* Chr.1: 35825749-35825749 on Build GRCh38
1667406 c.529G>T p.G177C Chr.1: 35825719-35825719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021573945 Chr.1: 35825814-35825814 on Build GRCh38
rs535953094 Chr.1: 35825715-35825715 on Build GRCh38
rs763394874 Chr.1: 35825711-35825711 on Build GRCh38
rs35950874 Chr.1: 35825716-35825717 on Build GRCh38
rs760085963 Chr.1: 35825899-35825900 on Build GRCh38
rs773146579 Chr.1: 35825856-35825856 on Build GRCh38
rs940936670 Chr.1: 35825870-35825870 on Build GRCh38
rs777168864 Chr.1: 35825739-35825739 on Build GRCh38
rs200471384 Chr.1: 35825788-35825788 on Build GRCh38
rs920280774 Chr.1: 35825881-35825881 on Build GRCh38
rs752305682 Chr.1: 35825718-35825718 on Build GRCh38
rs372001765 Chr.1: 35825840-35825840 on Build GRCh38
rs372002732 Chr.1: 35825894-35825894 on Build GRCh38
rs751952450 Chr.1: 35825888-35825889 on Build GRCh38
rs750905495 Chr.1: 35825710-35825710 on Build GRCh38
rs766770906 Chr.1: 35825858-35825859 on Build GRCh38
rs374925290 Chr.1: 35825716-35825716 on Build GRCh38
rs766990263 Chr.1: 35825877-35825877 on Build GRCh38
rs201828878 Chr.1: 35825795-35825795 on Build GRCh38
rs757152966 Chr.1: 35825811-35825811 on Build GRCh38
rs769872317 Chr.1: 35825853-35825853 on Build GRCh38
rs768024900 Chr.1: 35825886-35825886 on Build GRCh38
rs910414480 Chr.1: 35825871-35825871 on Build GRCh38
rs751943288 Chr.1: 35825714-35825714 on Build GRCh38
rs201060270 Chr.1: 35825824-35825824 on Build GRCh38
rs1012983873 Chr.1: 35825783-35825783 on Build GRCh38
rs761190583 Chr.1: 35825859-35825862 on Build GRCh38
rs750702347 Chr.1: 35825830-35825833 on Build GRCh38
rs765371293 Chr.1: 35825818-35825825 on Build GRCh38
rs778885830 Chr.1: 35825819-35825819 on Build GRCh38
rs767045147 Chr.1: 35825712-35825712 on Build GRCh38
rs759922975 Chr.1: 35825884-35825884 on Build GRCh38
rs755610082 Chr.1: 35825720-35825720 on Build GRCh38
rs375375625 Chr.1: 35825851-35825851 on Build GRCh38
rs200431448 Chr.1: 35825863-35825863 on Build GRCh38
rs547662402 Chr.1: 35825832-35825832 on Build GRCh38
rs755698629 Chr.1: 35825897-35825897 on Build GRCh38
rs368456437 Chr.1: 35825880-35825880 on Build GRCh38

View Full Product Details