Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87612114-87612377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSPP
Entrez Gene ID
1834
Gene Name
dentin sialophosphoprotein
Gene Aliases
DFNA39, DGI1, DMP3, DPP, DSP
UniGene
Hs.678914
Gene Chromosome Location
Chr.4: 87608529-87616873 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
256070 c.119C>G p.S40* Chr.4: 87612172-87612172 on Build GRCh38
4469426 c.159C>T p.T53T Chr.4: 87612345-87612345 on Build GRCh38
4126581 c.75G>A p.E25E Chr.4: 87612128-87612128 on Build GRCh38
3606839 c.119C>T p.S40L Chr.4: 87612172-87612172 on Build GRCh38
6649680 c.130G>T p.V44L Chr.4: 87612183-87612183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956897515 Chr.4: 87612155-87612155 on Build GRCh38
rs202163835 Chr.4: 87612144-87612144 on Build GRCh38
rs376270964 Chr.4: 87612334-87612334 on Build GRCh38
rs1050476689 Chr.4: 87612231-87612231 on Build GRCh38
rs752435946 Chr.4: 87612271-87612271 on Build GRCh38
rs121912985 Chr.4: 87612186-87612186 on Build GRCh38
rs539760943 Chr.4: 87612302-87612303 on Build GRCh38
rs748430202 Chr.4: 87612303-87612303 on Build GRCh38
rs766233158 Chr.4: 87612214-87612214 on Build GRCh38
rs199612946 Chr.4: 87612308-87612308 on Build GRCh38
rs13131929 Chr.4: 87612289-87612289 on Build GRCh38
rs562493366 Chr.4: 87612295-87612295 on Build GRCh38
rs925433833 Chr.4: 87612146-87612146 on Build GRCh38
rs760419956 Chr.4: 87612339-87612339 on Build GRCh38
rs758118247 Chr.4: 87612274-87612274 on Build GRCh38
rs923545944 Chr.4: 87612177-87612177 on Build GRCh38
rs758992859 Chr.4: 87612223-87612223 on Build GRCh38
rs767211266 Chr.4: 87612309-87612309 on Build GRCh38
rs760461844 Chr.4: 87612198-87612198 on Build GRCh38
rs768766639 Chr.4: 87612300-87612300 on Build GRCh38
rs1034004524 Chr.4: 87612273-87612273 on Build GRCh38
rs772873203 Chr.4: 87612166-87612166 on Build GRCh38
rs200641947 Chr.4: 87612138-87612138 on Build GRCh38
rs187578466 Chr.4: 87612330-87612330 on Build GRCh38
rs770712152 Chr.4: 87612340-87612340 on Build GRCh38
rs778963135 Chr.4: 87612302-87612302 on Build GRCh38
rs371832336 Chr.4: 87612222-87612222 on Build GRCh38
rs972753953 Chr.4: 87612352-87612352 on Build GRCh38
rs776482978 Chr.4: 87612342-87612342 on Build GRCh38
rs13131936 Chr.4: 87612293-87612293 on Build GRCh38
rs781728187 Chr.4: 87612288-87612288 on Build GRCh38
rs759513775 Chr.4: 87612357-87612357 on Build GRCh38
rs371905328 Chr.4: 87612137-87612137 on Build GRCh38
rs375217534 Chr.4: 87612238-87612238 on Build GRCh38

View Full Product Details