Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41585388-41585613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP4
Entrez Gene ID
116113
Gene Name
forkhead box P4
Gene Aliases
hFKHLA
UniGene
Hs.131436
Gene Chromosome Location
Chr.6: 41546426-41602384 on Build GRCh38
Transcripts
NM_001012426.1 NM_001012427.1 NM_138457.2 XM_006714991.3 XM_011514289.2 XM_011514290.2 XM_011514291.2 XM_011514292.2 XM_011514293.2 XM_017010233.1 XM_017010234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1721227 c.424-9delC p.? Chr.6: 41585422-41585422 on Build GRCh38
3949216 c.505A>G p.K169E Chr.6: 41585512-41585512 on Build GRCh38
5492072 c.424-9_424-8insC p.? Chr.6: 41585422-41585423 on Build GRCh38
5492071 c.424-9_424-8insC p.? Chr.6: 41585422-41585423 on Build GRCh38
3873977 c.429G>A p.Q143Q Chr.6: 41585436-41585436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765253728 Chr.6: 41585558-41585558 on Build GRCh38
rs771709521 Chr.6: 41585514-41585514 on Build GRCh38
rs1005568162 Chr.6: 41585470-41585470 on Build GRCh38
rs199622532 Chr.6: 41585447-41585447 on Build GRCh38
rs188320749 Chr.6: 41585554-41585554 on Build GRCh38
rs372409585 Chr.6: 41585546-41585546 on Build GRCh38
rs946436030 Chr.6: 41585570-41585570 on Build GRCh38
rs759986625 Chr.6: 41585427-41585427 on Build GRCh38
rs764167619 Chr.6: 41585550-41585550 on Build GRCh38
rs753175229 Chr.6: 41585428-41585428 on Build GRCh38
rs564668890 Chr.6: 41585586-41585586 on Build GRCh38
rs750575943 Chr.6: 41585559-41585559 on Build GRCh38
rs368450451 Chr.6: 41585545-41585545 on Build GRCh38
rs184723671 Chr.6: 41585425-41585425 on Build GRCh38
rs759989662 Chr.6: 41585540-41585540 on Build GRCh38
rs769729110 Chr.6: 41585417-41585417 on Build GRCh38
rs150633525 Chr.6: 41585504-41585504 on Build GRCh38
rs915006299 Chr.6: 41585560-41585560 on Build GRCh38
rs776142547 Chr.6: 41585446-41585447 on Build GRCh38
rs765968864 Chr.6: 41585422-41585422 on Build GRCh38
rs751794942 Chr.6: 41585460-41585460 on Build GRCh38
rs1003425135 Chr.6: 41585409-41585409 on Build GRCh38
rs369273150 Chr.6: 41585416-41585416 on Build GRCh38
rs775161768 Chr.6: 41585422-41585422 on Build GRCh38
rs780180861 Chr.6: 41585502-41585502 on Build GRCh38
rs200488844 Chr.6: 41585426-41585426 on Build GRCh38
rs996327233 Chr.6: 41585479-41585480 on Build GRCh38
rs770689434 Chr.6: 41585410-41585410 on Build GRCh38
rs760423295 Chr.6: 41585421-41585422 on Build GRCh38
rs868187232 Chr.6: 41585510-41585510 on Build GRCh38
rs758545129 Chr.6: 41585446-41585446 on Build GRCh38
rs773937574 Chr.6: 41585505-41585505 on Build GRCh38
rs759132444 Chr.6: 41585467-41585472 on Build GRCh38
rs148022939 Chr.6: 41585463-41585463 on Build GRCh38
rs369724251 Chr.6: 41585423-41585423 on Build GRCh38
rs767826727 Chr.6: 41585556-41585556 on Build GRCh38
rs780882681 Chr.6: 41585469-41585469 on Build GRCh38
rs368953290 Chr.6: 41585539-41585539 on Build GRCh38
rs755841725 Chr.6: 41585492-41585492 on Build GRCh38
rs977732161 Chr.6: 41585589-41585589 on Build GRCh38
rs141899153 Chr.6: 41585475-41585475 on Build GRCh38

View Full Product Details