Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 18659641-18659872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL5B
Entrez Gene ID
221079
Gene Name
ADP ribosylation factor like GTPase 5B
Gene Aliases
ARL8
UniGene
Hs.25362
Gene Chromosome Location
Chr.10: 18659335-18681639 on Build GRCh38
Transcripts
NM_178815.3 XM_005252400.1 XM_005252401.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5341216 c.31C>T p.L11F Chr.10: 18659668-18659668 on Build GRCh38
5036041 c.46+1G>T p.? Chr.10: 18659684-18659684 on Build GRCh38
6065538 c.38G>T p.C13F Chr.10: 18659675-18659675 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs959753387 Chr.10: 18659851-18659851 on Build GRCh38
rs976875294 Chr.10: 18659672-18659672 on Build GRCh38
rs768946901 Chr.10: 18659688-18659688 on Build GRCh38
rs753772963 Chr.10: 18659728-18659728 on Build GRCh38
rs1040985059 Chr.10: 18659762-18659762 on Build GRCh38
rs144696662 Chr.10: 18659661-18659661 on Build GRCh38
rs770265970 Chr.10: 18659670-18659670 on Build GRCh38
rs765621006 Chr.10: 18659687-18659689 on Build GRCh38
rs764803128 Chr.10: 18659745-18659745 on Build GRCh38
rs780532011 Chr.10: 18659680-18659680 on Build GRCh38
rs1029644890 Chr.10: 18659702-18659702 on Build GRCh38
rs573704740 Chr.10: 18659699-18659699 on Build GRCh38
rs1055296230 Chr.10: 18659769-18659769 on Build GRCh38
rs973666316 Chr.10: 18659734-18659734 on Build GRCh38
rs954030432 Chr.10: 18659707-18659707 on Build GRCh38
rs147434932 Chr.10: 18659846-18659846 on Build GRCh38
rs937686987 Chr.10: 18659765-18659765 on Build GRCh38
rs1042920663 Chr.10: 18659802-18659802 on Build GRCh38
rs986008828 Chr.10: 18659712-18659712 on Build GRCh38
rs894090261 Chr.10: 18659771-18659771 on Build GRCh38
rs1011057526 Chr.10: 18659793-18659793 on Build GRCh38
rs749654996 Chr.10: 18659682-18659682 on Build GRCh38
rs551599459 Chr.10: 18659792-18659792 on Build GRCh38
rs535004884 Chr.10: 18659735-18659735 on Build GRCh38
rs200270125 Chr.10: 18659721-18659721 on Build GRCh38
rs533008294 Chr.10: 18659741-18659741 on Build GRCh38
rs752060762 Chr.10: 18659843-18659843 on Build GRCh38
rs902697515 Chr.10: 18659834-18659834 on Build GRCh38
rs901993207 Chr.10: 18659774-18659774 on Build GRCh38
rs887794179 Chr.10: 18659829-18659829 on Build GRCh38
rs929020396 Chr.10: 18659747-18659747 on Build GRCh38
rs182696656 Chr.10: 18659799-18659799 on Build GRCh38
rs927596752 Chr.10: 18659760-18659760 on Build GRCh38
rs994186219 Chr.10: 18659781-18659781 on Build GRCh38
rs910937949 Chr.10: 18659746-18659746 on Build GRCh38
rs1046152154 Chr.10: 18659758-18659758 on Build GRCh38
rs111690788 Chr.10: 18659835-18659835 on Build GRCh38
rs760794178 Chr.10: 18659717-18659717 on Build GRCh38
rs540737636 Chr.10: 18659713-18659713 on Build GRCh38
rs943796790 Chr.10: 18659758-18659759 on Build GRCh38

View Full Product Details