Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73492517-73492766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf169
Entrez Gene ID
79697
Gene Name
chromosome 14 open reading frame 169
Gene Aliases
MAPJD, NO66, ROX, URLC2, hsNO66
UniGene
Hs.509916
Gene Chromosome Location
Chr.14: 73490934-73493401 on Build GRCh38
Transcripts
NM_024644.4 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_017021289.1

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
UniGene
Hs.660774
Gene Chromosome Location
Chr.14: 73478484-73634418 on Build GRCh38
Transcripts
NM_024644.4 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_017021289.1

SNP Detail

SNP ID Chromosome Location
rs773233651 Chr.14: 73492697-73492699 on Build GRCh38
rs758940009 Chr.14: 73492595-73492595 on Build GRCh38
rs751393987 Chr.14: 73492702-73492702 on Build GRCh38
rs762945872 Chr.14: 73492715-73492718 on Build GRCh38
rs373711592 Chr.14: 73492720-73492720 on Build GRCh38
rs764991790 Chr.14: 73492603-73492603 on Build GRCh38
rs113610134 Chr.14: 73492701-73492701 on Build GRCh38
rs770092953 Chr.14: 73492538-73492538 on Build GRCh38
rs762536716 Chr.14: 73492697-73492697 on Build GRCh38
rs759147551 Chr.14: 73492686-73492686 on Build GRCh38
rs769071468 Chr.14: 73492595-73492599 on Build GRCh38
rs780386713 Chr.14: 73492639-73492639 on Build GRCh38
rs774998626 Chr.14: 73492580-73492580 on Build GRCh38
rs34068039 Chr.14: 73492604-73492604 on Build GRCh38
rs775214366 Chr.14: 73492695-73492695 on Build GRCh38
rs748415685 Chr.14: 73492684-73492684 on Build GRCh38
rs377065746 Chr.14: 73492667-73492667 on Build GRCh38
rs756218398 Chr.14: 73492724-73492724 on Build GRCh38
rs370357833 Chr.14: 73492712-73492712 on Build GRCh38
rs535810576 Chr.14: 73492631-73492631 on Build GRCh38
rs373839917 Chr.14: 73492659-73492659 on Build GRCh38
rs1003703772 Chr.14: 73492668-73492668 on Build GRCh38
rs763810203 Chr.14: 73492605-73492605 on Build GRCh38
rs754132267 Chr.14: 73492737-73492737 on Build GRCh38
rs749452153 Chr.14: 73492643-73492643 on Build GRCh38
rs576690817 Chr.14: 73492730-73492730 on Build GRCh38
rs770023747 Chr.14: 73492693-73492699 on Build GRCh38
rs764651514 Chr.14: 73492693-73492693 on Build GRCh38
rs749715987 Chr.14: 73492540-73492540 on Build GRCh38
rs748700741 Chr.14: 73492663-73492663 on Build GRCh38
rs370838005 Chr.14: 73492636-73492636 on Build GRCh38
rs376251702 Chr.14: 73492620-73492620 on Build GRCh38
rs200159534 Chr.14: 73492589-73492589 on Build GRCh38
rs781357389 Chr.14: 73492639-73492639 on Build GRCh38
rs376001064 Chr.14: 73492577-73492577 on Build GRCh38
rs775552159 Chr.14: 73492539-73492539 on Build GRCh38
rs773572459 Chr.14: 73492675-73492675 on Build GRCh38
rs780960346 Chr.14: 73492629-73492629 on Build GRCh38
rs766012728 Chr.14: 73492592-73492592 on Build GRCh38
rs761573614 Chr.14: 73492703-73492703 on Build GRCh38
rs757144885 Chr.14: 73492628-73492628 on Build GRCh38
rs556655407 Chr.14: 73492647-73492647 on Build GRCh38
rs1036502932 Chr.14: 73492727-73492727 on Build GRCh38
rs776171171 Chr.14: 73492593-73492593 on Build GRCh38

View Full Product Details