Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 154121901-154122158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF26
Entrez Gene ID
26084
Gene Name
Rho guanine nucleotide exchange factor 26
Gene Aliases
CSGEF, HMFN1864, SGEF
UniGene
Hs.240845
Gene Chromosome Location
Chr.3: 154121003-154257827 on Build GRCh38
Transcripts
NM_001251962.1 NM_001251963.1 NM_015595.3 XM_011512672.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3004113 c.21G>A p.V7V Chr.3: 154122013-154122013 on Build GRCh38
3004121 c.86T>G p.V29G Chr.3: 154122078-154122078 on Build GRCh38
5059419 c.130A>G p.N44D Chr.3: 154122122-154122122 on Build GRCh38
5521047 c.70C>T p.P24S Chr.3: 154122062-154122062 on Build GRCh38
4934605 c.86T>C p.V29A Chr.3: 154122078-154122078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763284937 Chr.3: 154121953-154121953 on Build GRCh38
rs745450549 Chr.3: 154122030-154122030 on Build GRCh38
rs755382272 Chr.3: 154122052-154122052 on Build GRCh38
rs763940482 Chr.3: 154122023-154122023 on Build GRCh38
rs771323300 Chr.3: 154121948-154121966 on Build GRCh38
rs199795997 Chr.3: 154121962-154121962 on Build GRCh38
rs200926651 Chr.3: 154122078-154122078 on Build GRCh38
rs763048960 Chr.3: 154122010-154122010 on Build GRCh38
rs374549416 Chr.3: 154122007-154122007 on Build GRCh38
rs757938558 Chr.3: 154122038-154122038 on Build GRCh38
rs756714768 Chr.3: 154121978-154121978 on Build GRCh38
rs769226137 Chr.3: 154122001-154122001 on Build GRCh38
rs890249420 Chr.3: 154122116-154122116 on Build GRCh38
rs959638150 Chr.3: 154122084-154122084 on Build GRCh38
rs771077073 Chr.3: 154122066-154122066 on Build GRCh38
rs915470090 Chr.3: 154122097-154122097 on Build GRCh38
rs780524621 Chr.3: 154121992-154121992 on Build GRCh38
rs757312479 Chr.3: 154122131-154122131 on Build GRCh38
rs760559763 Chr.3: 154121963-154121963 on Build GRCh38
rs200140877 Chr.3: 154122014-154122014 on Build GRCh38
rs747398863 Chr.3: 154121993-154121993 on Build GRCh38
rs1042514109 Chr.3: 154122124-154122124 on Build GRCh38
rs760465611 Chr.3: 154122021-154122021 on Build GRCh38
rs1042565708 Chr.3: 154121968-154121968 on Build GRCh38
rs367824965 Chr.3: 154121984-154121984 on Build GRCh38
rs927978539 Chr.3: 154121933-154121933 on Build GRCh38
rs374318138 Chr.3: 154122125-154122125 on Build GRCh38
rs774549332 Chr.3: 154122076-154122076 on Build GRCh38
rs372226090 Chr.3: 154121987-154121987 on Build GRCh38
rs1051695742 Chr.3: 154122112-154122112 on Build GRCh38
rs375842704 Chr.3: 154122083-154122083 on Build GRCh38
rs748748466 Chr.3: 154122004-154122004 on Build GRCh38
rs12493885 Chr.3: 154122077-154122077 on Build GRCh38
rs374286801 Chr.3: 154121982-154121982 on Build GRCh38
rs552873638 Chr.3: 154122119-154122119 on Build GRCh38
rs188383913 Chr.3: 154122063-154122063 on Build GRCh38
rs776525028 Chr.3: 154122081-154122081 on Build GRCh38
rs751325925 Chr.3: 154122049-154122049 on Build GRCh38
rs990874567 Chr.3: 154122086-154122086 on Build GRCh38
rs915372962 Chr.3: 154121954-154121954 on Build GRCh38
rs748571817 Chr.3: 154122061-154122061 on Build GRCh38
rs754718776 Chr.3: 154122109-154122109 on Build GRCh38
rs541643715 Chr.3: 154122028-154122028 on Build GRCh38
rs369109073 Chr.3: 154122123-154122123 on Build GRCh38
rs773917501 Chr.3: 154121967-154121967 on Build GRCh38
rs563109480 Chr.3: 154122048-154122048 on Build GRCh38
rs767189484 Chr.3: 154121956-154121956 on Build GRCh38
rs746054521 Chr.3: 154122140-154122140 on Build GRCh38
rs764034579 Chr.3: 154121964-154121964 on Build GRCh38
rs781724116 Chr.3: 154122058-154122058 on Build GRCh38
rs994667023 Chr.3: 154122057-154122057 on Build GRCh38
rs779010194 Chr.3: 154122138-154122138 on Build GRCh38
rs368976915 Chr.3: 154122079-154122079 on Build GRCh38
rs753275488 Chr.3: 154121967-154121967 on Build GRCh38
rs982096151 Chr.3: 154121929-154121929 on Build GRCh38
rs761615412 Chr.3: 154122082-154122082 on Build GRCh38
rs749402094 Chr.3: 154122064-154122064 on Build GRCh38
rs191718226 Chr.3: 154122128-154122128 on Build GRCh38
rs201682467 Chr.3: 154121952-154121952 on Build GRCh38
rs753759594 Chr.3: 154122025-154122025 on Build GRCh38
rs751235813 Chr.3: 154122105-154122105 on Build GRCh38
rs941455212 Chr.3: 154122070-154122070 on Build GRCh38
rs772537950 Chr.3: 154122087-154122087 on Build GRCh38
rs761855865 Chr.3: 154122026-154122026 on Build GRCh38
rs199680317 Chr.3: 154121948-154121948 on Build GRCh38
rs759243845 Chr.3: 154122128-154122130 on Build GRCh38
rs139430962 Chr.3: 154122017-154122017 on Build GRCh38
rs938247762 Chr.3: 154121950-154121950 on Build GRCh38
rs528513610 Chr.3: 154122088-154122088 on Build GRCh38
rs756680473 Chr.3: 154122062-154122062 on Build GRCh38

View Full Product Details