Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 143495646-143495915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUCA2
Entrez Gene ID
2519
Gene Name
fucosidase, alpha-L- 2, plasma
Gene Aliases
dJ20N2.5
UniGene
Hs.17680
Gene Chromosome Location
Chr.6: 143494812-143511883 on Build GRCh38
Transcripts
NM_032020.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6297567 c.1354C>A p.Q452K Chr.6: 143495757-143495757 on Build GRCh38
1074053 c.1287G>A p.Q429Q Chr.6: 143495824-143495824 on Build GRCh38
6430612 c.1317T>A p.N439K Chr.6: 143495794-143495794 on Build GRCh38
3829193 c.1271T>G p.L424R Chr.6: 143495840-143495840 on Build GRCh38
6413527 c.1289C>A p.P430Q Chr.6: 143495822-143495822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754694312 Chr.6: 143495808-143495808 on Build GRCh38
rs779526135 Chr.6: 143495730-143495730 on Build GRCh38
rs150307909 Chr.6: 143495786-143495786 on Build GRCh38
rs865900342 Chr.6: 143495834-143495834 on Build GRCh38
rs758253790 Chr.6: 143495696-143495696 on Build GRCh38
rs767233296 Chr.6: 143495838-143495838 on Build GRCh38
rs765270351 Chr.6: 143495773-143495773 on Build GRCh38
rs34274568 Chr.6: 143495691-143495692 on Build GRCh38
rs761751116 Chr.6: 143495676-143495676 on Build GRCh38
rs200632280 Chr.6: 143495750-143495750 on Build GRCh38
rs899761799 Chr.6: 143495794-143495794 on Build GRCh38
rs151156975 Chr.6: 143495732-143495732 on Build GRCh38
rs777129921 Chr.6: 143495872-143495872 on Build GRCh38
rs991244160 Chr.6: 143495807-143495807 on Build GRCh38
rs775711590 Chr.6: 143495733-143495733 on Build GRCh38
rs549463275 Chr.6: 143495783-143495783 on Build GRCh38
rs8161 Chr.6: 143495722-143495722 on Build GRCh38
rs750214725 Chr.6: 143495688-143495688 on Build GRCh38
rs750424478 Chr.6: 143495776-143495776 on Build GRCh38
rs1030197247 Chr.6: 143495703-143495703 on Build GRCh38
rs780855602 Chr.6: 143495716-143495716 on Build GRCh38
rs368918467 Chr.6: 143495706-143495706 on Build GRCh38
rs752290485 Chr.6: 143495841-143495841 on Build GRCh38
rs201420341 Chr.6: 143495726-143495726 on Build GRCh38
rs78275140 Chr.6: 143495779-143495779 on Build GRCh38
rs140243222 Chr.6: 143495749-143495749 on Build GRCh38
rs758834187 Chr.6: 143495782-143495783 on Build GRCh38
rs138290108 Chr.6: 143495787-143495787 on Build GRCh38
rs755729641 Chr.6: 143495862-143495862 on Build GRCh38
rs765109071 Chr.6: 143495685-143495685 on Build GRCh38
rs746335288 Chr.6: 143495731-143495731 on Build GRCh38
rs779691200 Chr.6: 143495705-143495705 on Build GRCh38
rs754674124 Chr.6: 143495708-143495708 on Build GRCh38

View Full Product Details