Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162277538-162277763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFIH1
Entrez Gene ID
64135
Gene Name
interferon induced with helicase C domain 1
Gene Aliases
AGS7, Hlcd, IDDM19, MDA-5, MDA5, RLR-2, SGMRT1
UniGene
Hs.163173
Gene Chromosome Location
Chr.2: 162267079-162318708 on Build GRCh38
Transcripts
NM_022168.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
207537 c.1865C>T p.A622V Chr.2: 162277594-162277594 on Build GRCh38
1285470 c.1843G>A p.D615N Chr.2: 162277616-162277616 on Build GRCh38
3971664 c.1913T>G p.F638C Chr.2: 162277546-162277546 on Build GRCh38
6509906 c.1804G>T p.E602* Chr.2: 162277655-162277655 on Build GRCh38
395212 c.1907A>C p.K636T Chr.2: 162277552-162277552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375482697 Chr.2: 162277604-162277604 on Build GRCh38
rs200017837 Chr.2: 162277606-162277606 on Build GRCh38
rs745937740 Chr.2: 162277607-162277607 on Build GRCh38
rs200980825 Chr.2: 162277573-162277573 on Build GRCh38
rs963903258 Chr.2: 162277670-162277670 on Build GRCh38
rs759122723 Chr.2: 162277664-162277664 on Build GRCh38
rs35744605 Chr.2: 162277580-162277580 on Build GRCh38
rs755951360 Chr.2: 162277672-162277672 on Build GRCh38
rs772083054 Chr.2: 162277621-162277621 on Build GRCh38
rs768993250 Chr.2: 162277690-162277690 on Build GRCh38
rs749942019 Chr.2: 162277586-162277586 on Build GRCh38
rs145818112 Chr.2: 162277574-162277574 on Build GRCh38
rs558387118 Chr.2: 162277631-162277631 on Build GRCh38
rs145792185 Chr.2: 162277693-162277693 on Build GRCh38
rs935247878 Chr.2: 162277711-162277711 on Build GRCh38
rs780212949 Chr.2: 162277625-162277625 on Build GRCh38
rs770016404 Chr.2: 162277707-162277707 on Build GRCh38
rs1055824554 Chr.2: 162277663-162277663 on Build GRCh38
rs765761238 Chr.2: 162277658-162277658 on Build GRCh38
rs762331564 Chr.2: 162277637-162277637 on Build GRCh38
rs200945986 Chr.2: 162277666-162277666 on Build GRCh38
rs763549486 Chr.2: 162277653-162277653 on Build GRCh38
rs118172671 Chr.2: 162277647-162277647 on Build GRCh38
rs773729704 Chr.2: 162277731-162277731 on Build GRCh38
rs74162084 Chr.2: 162277675-162277675 on Build GRCh38
rs763314815 Chr.2: 162277712-162277712 on Build GRCh38
rs562561591 Chr.2: 162277738-162277738 on Build GRCh38
rs1025854546 Chr.2: 162277685-162277685 on Build GRCh38
rs766027130 Chr.2: 162277593-162277593 on Build GRCh38
rs186942719 Chr.2: 162277594-162277594 on Build GRCh38
rs183649893 Chr.2: 162277696-162277696 on Build GRCh38
rs545610093 Chr.2: 162277576-162277576 on Build GRCh38
rs769009487 Chr.2: 162277632-162277632 on Build GRCh38
rs777064938 Chr.2: 162277636-162277636 on Build GRCh38
rs199818504 Chr.2: 162277571-162277571 on Build GRCh38
rs548261817 Chr.2: 162277698-162277699 on Build GRCh38
rs766039450 Chr.2: 162277662-162277672 on Build GRCh38
rs775623826 Chr.2: 162277570-162277570 on Build GRCh38
rs767136509 Chr.2: 162277740-162277740 on Build GRCh38
rs771474528 Chr.2: 162277723-162277723 on Build GRCh38
rs773795297 Chr.2: 162277664-162277664 on Build GRCh38
rs762422699 Chr.2: 162277587-162277587 on Build GRCh38
rs374369372 Chr.2: 162277708-162277708 on Build GRCh38
rs200741709 Chr.2: 162277667-162277667 on Build GRCh38
rs747086201 Chr.2: 162277686-162277686 on Build GRCh38
rs752494417 Chr.2: 162277599-162277599 on Build GRCh38
rs191839015 Chr.2: 162277676-162277676 on Build GRCh38
rs768024482 Chr.2: 162277699-162277699 on Build GRCh38
rs770726979 Chr.2: 162277597-162277597 on Build GRCh38
rs751095720 Chr.2: 162277662-162277662 on Build GRCh38
rs773635540 Chr.2: 162277648-162277648 on Build GRCh38

View Full Product Details