Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 216058824-216059055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PECR
Entrez Gene ID
55825
Gene Name
peroxisomal trans-2-enoyl-CoA reductase
Gene Aliases
DCRRP, HPDHASE, HSA250303, PVIARL, SDR29C1, TERP
UniGene
Hs.281680
Gene Chromosome Location
Chr.2: 216029088-216081823 on Build GRCh38
Transcripts
NM_018441.5 XM_011511478.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
573618 c.478C>T p.P160S Chr.2: 216058923-216058923 on Build GRCh38
4922319 c.498A>T p.P166P Chr.2: 216058903-216058903 on Build GRCh38
3577559 c.477C>T p.V159V Chr.2: 216058924-216058924 on Build GRCh38
5491572 c.449A>G p.H150R Chr.2: 216058952-216058952 on Build GRCh38
7215104 c.445G>A p.E149K Chr.2: 216058956-216058956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778405825 Chr.2: 216059013-216059013 on Build GRCh38
rs756053288 Chr.2: 216059001-216059001 on Build GRCh38
rs746777056 Chr.2: 216058882-216058882 on Build GRCh38
rs141695178 Chr.2: 216058961-216058961 on Build GRCh38
rs745756389 Chr.2: 216058889-216058889 on Build GRCh38
rs748143243 Chr.2: 216058980-216058980 on Build GRCh38
rs772035471 Chr.2: 216059030-216059030 on Build GRCh38
rs201488757 Chr.2: 216058992-216058992 on Build GRCh38
rs746674353 Chr.2: 216059016-216059017 on Build GRCh38
rs745801486 Chr.2: 216058997-216058997 on Build GRCh38
rs768061100 Chr.2: 216058925-216058925 on Build GRCh38
rs768177191 Chr.2: 216058905-216058905 on Build GRCh38
rs777683775 Chr.2: 216058880-216058880 on Build GRCh38
rs369859130 Chr.2: 216059012-216059012 on Build GRCh38
rs774844192 Chr.2: 216058920-216058920 on Build GRCh38
rs762132068 Chr.2: 216058921-216058921 on Build GRCh38
rs748981310 Chr.2: 216058904-216058904 on Build GRCh38
rs768393421 Chr.2: 216059008-216059008 on Build GRCh38
rs774791648 Chr.2: 216058898-216058898 on Build GRCh38
rs757304362 Chr.2: 216058883-216058883 on Build GRCh38
rs779670167 Chr.2: 216059003-216059003 on Build GRCh38
rs773730447 Chr.2: 216058934-216058934 on Build GRCh38
rs748741044 Chr.2: 216059004-216059004 on Build GRCh38
rs772231905 Chr.2: 216059031-216059031 on Build GRCh38
rs374436429 Chr.2: 216058939-216058939 on Build GRCh38
rs138408075 Chr.2: 216058938-216058938 on Build GRCh38
rs1034186599 Chr.2: 216058935-216058935 on Build GRCh38
rs373409927 Chr.2: 216059020-216059020 on Build GRCh38
rs62001895 Chr.2: 216058896-216058896 on Build GRCh38
rs983809299 Chr.2: 216059024-216059024 on Build GRCh38
rs1429148 Chr.2: 216058956-216058956 on Build GRCh38
rs755295078 Chr.2: 216058952-216058952 on Build GRCh38
rs781098611 Chr.2: 216058985-216058985 on Build GRCh38
rs772984269 Chr.2: 216059036-216059036 on Build GRCh38
rs200329052 Chr.2: 216058888-216058888 on Build GRCh38
rs757145058 Chr.2: 216058982-216058982 on Build GRCh38

View Full Product Details