Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 24587120-24587387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCYT1B
Entrez Gene ID
9468
Gene Name
phosphate cytidylyltransferase 1, choline, beta
Gene Aliases
CCTB, CTB
UniGene
Hs.660708
Gene Chromosome Location
Chr.X: 24558087-24672862 on Build GRCh38
Transcripts
NM_001163264.1 NM_001163265.1 NM_004845.4 XM_017029977.1

SNP Detail

SNP ID Chromosome Location
rs879004885 Chr.X: 24587196-24587196 on Build GRCh38
rs770128567 Chr.X: 24587257-24587257 on Build GRCh38
rs371475250 Chr.X: 24587340-24587340 on Build GRCh38
rs759576974 Chr.X: 24587333-24587333 on Build GRCh38
rs779940280 Chr.X: 24587209-24587209 on Build GRCh38
rs1055809607 Chr.X: 24587217-24587217 on Build GRCh38
rs772981585 Chr.X: 24587282-24587282 on Build GRCh38
rs750985354 Chr.X: 24587295-24587295 on Build GRCh38
rs747752357 Chr.X: 24587327-24587328 on Build GRCh38
rs767601378 Chr.X: 24587367-24587367 on Build GRCh38
rs184397456 Chr.X: 24587161-24587161 on Build GRCh38
rs142421826 Chr.X: 24587210-24587210 on Build GRCh38
rs1042492488 Chr.X: 24587228-24587228 on Build GRCh38
rs769116156 Chr.X: 24587221-24587221 on Build GRCh38
rs954362758 Chr.X: 24587146-24587146 on Build GRCh38
rs186666095 Chr.X: 24587230-24587230 on Build GRCh38
rs1055408603 Chr.X: 24587328-24587328 on Build GRCh38
rs111304630 Chr.X: 24587192-24587192 on Build GRCh38
rs148794543 Chr.X: 24587169-24587169 on Build GRCh38
rs34701484 Chr.X: 24587290-24587290 on Build GRCh38
rs762217678 Chr.X: 24587233-24587233 on Build GRCh38
rs961961292 Chr.X: 24587174-24587174 on Build GRCh38
rs746074014 Chr.X: 24587202-24587202 on Build GRCh38
rs201631061 Chr.X: 24587204-24587204 on Build GRCh38
rs755940545 Chr.X: 24587191-24587191 on Build GRCh38
rs976604135 Chr.X: 24587156-24587156 on Build GRCh38

View Full Product Details