Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30377613-30377856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYLPF
Entrez Gene ID
29895
Gene Name
myosin light chain, phosphorylatable, fast skeletal muscle
Gene Aliases
HUMMLC2B, MLC2B, MRLC2, MYL11
UniGene
Hs.50889
Gene Chromosome Location
Chr.16: 30370934-30377991 on Build GRCh38
Transcripts
NM_001324458.1 NM_001324459.1 NM_013292.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3420934 c.411G>T p.K137N Chr.16: 30377801-30377801 on Build GRCh38
344431 c.364G>A p.E122K Chr.16: 30377661-30377661 on Build GRCh38
4704746 c.429delC p.D146fs*>24 Chr.16: 30377819-30377819 on Build GRCh38
4856852 c.372G>A p.L124L Chr.16: 30377669-30377669 on Build GRCh38
5706103 c.424G>A p.A142T Chr.16: 30377814-30377814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024980456 Chr.16: 30377743-30377743 on Build GRCh38
rs751361323 Chr.16: 30377766-30377766 on Build GRCh38
rs758681075 Chr.16: 30377758-30377758 on Build GRCh38
rs777026825 Chr.16: 30377819-30377819 on Build GRCh38
rs761160620 Chr.16: 30377776-30377776 on Build GRCh38
rs201616986 Chr.16: 30377747-30377747 on Build GRCh38
rs1031881720 Chr.16: 30377659-30377659 on Build GRCh38
rs201069396 Chr.16: 30377749-30377749 on Build GRCh38
rs775600240 Chr.16: 30377764-30377764 on Build GRCh38
rs754711638 Chr.16: 30377767-30377767 on Build GRCh38
rs747388501 Chr.16: 30377801-30377801 on Build GRCh38
rs571468024 Chr.16: 30377730-30377730 on Build GRCh38
rs764699750 Chr.16: 30377739-30377739 on Build GRCh38
rs774078061 Chr.16: 30377799-30377799 on Build GRCh38
rs386352335 Chr.16: 30377821-30377821 on Build GRCh38
rs768849724 Chr.16: 30377806-30377806 on Build GRCh38
rs117563518 Chr.16: 30377787-30377787 on Build GRCh38
rs1047139818 Chr.16: 30377775-30377775 on Build GRCh38
rs748792726 Chr.16: 30377790-30377790 on Build GRCh38
rs747707614 Chr.16: 30377669-30377669 on Build GRCh38
rs780925693 Chr.16: 30377658-30377658 on Build GRCh38
rs774741708 Chr.16: 30377823-30377823 on Build GRCh38
rs747995265 Chr.16: 30377777-30377777 on Build GRCh38
rs946196843 Chr.16: 30377653-30377653 on Build GRCh38
rs957661958 Chr.16: 30377813-30377813 on Build GRCh38
rs770684007 Chr.16: 30377718-30377718 on Build GRCh38
rs781212632 Chr.16: 30377776-30377776 on Build GRCh38
rs764654468 Chr.16: 30377807-30377828 on Build GRCh38
rs1031975596 Chr.16: 30377805-30377805 on Build GRCh38
rs369244674 Chr.16: 30377824-30377824 on Build GRCh38
rs762733493 Chr.16: 30377716-30377716 on Build GRCh38
rs756468555 Chr.16: 30377663-30377663 on Build GRCh38
rs142749402 Chr.16: 30377792-30377792 on Build GRCh38
rs780252260 Chr.16: 30377760-30377760 on Build GRCh38
rs772948775 Chr.16: 30377715-30377715 on Build GRCh38
rs769137763 Chr.16: 30377696-30377696 on Build GRCh38
rs754372271 Chr.16: 30377818-30377819 on Build GRCh38
rs754237909 Chr.16: 30377741-30377741 on Build GRCh38
rs868443059 Chr.16: 30377818-30377818 on Build GRCh38
rs1019423134 Chr.16: 30377782-30377782 on Build GRCh38
rs538748459 Chr.16: 30377733-30377733 on Build GRCh38
rs767557906 Chr.16: 30377751-30377760 on Build GRCh38
rs867095046 Chr.16: 30377745-30377745 on Build GRCh38
rs369183798 Chr.16: 30377830-30377830 on Build GRCh38
rs368767733 Chr.16: 30377755-30377755 on Build GRCh38
rs371915471 Chr.16: 30377784-30377784 on Build GRCh38

View Full Product Details