Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65713725-65713950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT5
Entrez Gene ID
10524
Gene Name
lysine acetyltransferase 5
Gene Aliases
ESA1, HTATIP, HTATIP1, PLIP, TIP, TIP60, ZC2HC5, cPLA2
UniGene
Hs.397010
Gene Chromosome Location
Chr.11: 65711996-65719606 on Build GRCh38
Transcripts
NM_001206833.1 NM_006388.3 NM_182709.2 NM_182710.2 XM_006718421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6261554 c.648G>T p.Q216H Chr.11: 65713806-65713806 on Build GRCh38
5418301 c.690+9G>T p.? Chr.11: 65713857-65713857 on Build GRCh38
4165906 c.662G>C p.R221P Chr.11: 65713820-65713820 on Build GRCh38
4620642 c.655C>T p.R219W Chr.11: 65713813-65713813 on Build GRCh38
7202204 c.669G>A p.S223S Chr.11: 65713827-65713827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746852848 Chr.11: 65713764-65713764 on Build GRCh38
rs41302415 Chr.11: 65713758-65713758 on Build GRCh38
rs386834229 Chr.11: 65713796-65713796 on Build GRCh38
rs748829005 Chr.11: 65713873-65713873 on Build GRCh38
rs143413716 Chr.11: 65713781-65713781 on Build GRCh38
rs768864155 Chr.11: 65713783-65713783 on Build GRCh38
rs760305548 Chr.11: 65713813-65713813 on Build GRCh38
rs769402320 Chr.11: 65713776-65713776 on Build GRCh38
rs140269437 Chr.11: 65713867-65713867 on Build GRCh38
rs745363293 Chr.11: 65713860-65713860 on Build GRCh38
rs757179191 Chr.11: 65713852-65713852 on Build GRCh38
rs756391565 Chr.11: 65713746-65713746 on Build GRCh38
rs755576862 Chr.11: 65713861-65713861 on Build GRCh38
rs201879725 Chr.11: 65713827-65713827 on Build GRCh38
rs935809328 Chr.11: 65713920-65713920 on Build GRCh38
rs1016618224 Chr.11: 65713774-65713774 on Build GRCh38
rs774568647 Chr.11: 65713887-65713887 on Build GRCh38
rs762353780 Chr.11: 65713795-65713795 on Build GRCh38
rs769080127 Chr.11: 65713879-65713879 on Build GRCh38
rs911516666 Chr.11: 65713831-65713831 on Build GRCh38
rs529240661 Chr.11: 65713826-65713826 on Build GRCh38
rs562928893 Chr.11: 65713832-65713832 on Build GRCh38
rs780717977 Chr.11: 65713855-65713855 on Build GRCh38
rs376433951 Chr.11: 65713768-65713768 on Build GRCh38
rs773422520 Chr.11: 65713798-65713798 on Build GRCh38
rs758079719 Chr.11: 65713753-65713753 on Build GRCh38
rs767764622 Chr.11: 65713797-65713797 on Build GRCh38
rs540553489 Chr.11: 65713782-65713782 on Build GRCh38
rs767323485 Chr.11: 65713861-65713863 on Build GRCh38
rs372880133 Chr.11: 65713814-65713814 on Build GRCh38
rs71455782 Chr.11: 65713905-65713905 on Build GRCh38
rs560560349 Chr.11: 65713807-65713807 on Build GRCh38
rs753965581 Chr.11: 65713811-65713811 on Build GRCh38
rs780664854 Chr.11: 65713769-65713769 on Build GRCh38
rs571340239 Chr.11: 65713907-65713907 on Build GRCh38
rs773560498 Chr.11: 65713866-65713866 on Build GRCh38
rs370475272 Chr.11: 65713770-65713770 on Build GRCh38
rs962863089 Chr.11: 65713749-65713749 on Build GRCh38
rs369137034 Chr.11: 65713802-65713802 on Build GRCh38
rs752514360 Chr.11: 65713750-65713750 on Build GRCh38
rs573766757 Chr.11: 65713748-65713748 on Build GRCh38
rs935970611 Chr.11: 65713834-65713834 on Build GRCh38
rs765009426 Chr.11: 65713748-65713748 on Build GRCh38
rs111779388 Chr.11: 65713850-65713850 on Build GRCh38
rs939214431 Chr.11: 65713908-65713908 on Build GRCh38

View Full Product Details