Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50090383-50090620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MOV10L1
Entrez Gene ID
54456
Gene Name
Mov10 RISC complex RNA helicase like 1
Gene Aliases
CHAMP, DJ402G11.8
UniGene
Hs.62880
Gene Chromosome Location
Chr.22: 50090006-50161705 on Build GRCh38
Transcripts
NM_001164104.1 NM_001164105.1 NM_018995.2 XM_005261923.3 XM_011530696.1 XM_011530697.1 XM_011530698.1 XM_011530699.1 XM_011530701.1 XM_011530702.1 XM_011530704.1 XM_017028833.1 XM_017028834.1 XM_017028837.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5426245 c.12C>T p.L4L Chr.22: 50090487-50090487 on Build GRCh38
5834212 c.16delG p.V6fs*12 Chr.22: 50090491-50090491 on Build GRCh38
5591883 c.23G>A p.S8N Chr.22: 50090498-50090498 on Build GRCh38
1035302 c.21C>T p.R7R Chr.22: 50090496-50090496 on Build GRCh38
6714515 c.25G>A p.V9I Chr.22: 50090500-50090500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945472019 Chr.22: 50090577-50090577 on Build GRCh38
rs778617060 Chr.22: 50090518-50090519 on Build GRCh38
rs774575066 Chr.22: 50090428-50090428 on Build GRCh38
rs776842362 Chr.22: 50090507-50090507 on Build GRCh38
rs372611802 Chr.22: 50090485-50090485 on Build GRCh38
rs997383029 Chr.22: 50090579-50090579 on Build GRCh38
rs772445427 Chr.22: 50090494-50090494 on Build GRCh38
rs1000690600 Chr.22: 50090414-50090414 on Build GRCh38
rs768911277 Chr.22: 50090501-50090501 on Build GRCh38
rs961430833 Chr.22: 50090519-50090519 on Build GRCh38
rs969980343 Chr.22: 50090403-50090403 on Build GRCh38
rs756041523 Chr.22: 50090531-50090531 on Build GRCh38
rs750062613 Chr.22: 50090546-50090546 on Build GRCh38
rs913960087 Chr.22: 50090571-50090571 on Build GRCh38
rs141552799 Chr.22: 50090511-50090512 on Build GRCh38
rs181326638 Chr.22: 50090404-50090404 on Build GRCh38
rs149871903 Chr.22: 50090545-50090545 on Build GRCh38
rs546507444 Chr.22: 50090418-50090418 on Build GRCh38
rs758887431 Chr.22: 50090547-50090547 on Build GRCh38
rs781649847 Chr.22: 50090582-50090582 on Build GRCh38
rs776094426 Chr.22: 50090431-50090431 on Build GRCh38
rs772472452 Chr.22: 50090429-50090429 on Build GRCh38
rs763952958 Chr.22: 50090538-50090538 on Build GRCh38
rs777252001 Chr.22: 50090467-50090467 on Build GRCh38
rs1049873216 Chr.22: 50090475-50090475 on Build GRCh38
rs1053915189 Chr.22: 50090422-50090422 on Build GRCh38
rs749008365 Chr.22: 50090477-50090477 on Build GRCh38
rs140007445 Chr.22: 50090405-50090405 on Build GRCh38
rs776183432 Chr.22: 50090495-50090495 on Build GRCh38
rs556967077 Chr.22: 50090550-50090550 on Build GRCh38
rs765389529 Chr.22: 50090511-50090511 on Build GRCh38
rs552522724 Chr.22: 50090500-50090500 on Build GRCh38
rs570793867 Chr.22: 50090508-50090508 on Build GRCh38
rs769287580 Chr.22: 50090551-50090551 on Build GRCh38
rs371047044 Chr.22: 50090560-50090560 on Build GRCh38
rs773433665 Chr.22: 50090512-50090512 on Build GRCh38
rs755660781 Chr.22: 50090464-50090464 on Build GRCh38
rs747333113 Chr.22: 50090530-50090530 on Build GRCh38
rs143891230 Chr.22: 50090548-50090548 on Build GRCh38
rs747698593 Chr.22: 50090537-50090538 on Build GRCh38
rs759279711 Chr.22: 50090522-50090522 on Build GRCh38
rs544684845 Chr.22: 50090480-50090480 on Build GRCh38
rs761924070 Chr.22: 50090437-50090437 on Build GRCh38
rs771344998 Chr.22: 50090573-50090573 on Build GRCh38
rs764241935 Chr.22: 50090434-50090434 on Build GRCh38
rs529417188 Chr.22: 50090457-50090457 on Build GRCh38
rs760871485 Chr.22: 50090433-50090433 on Build GRCh38
rs1030301664 Chr.22: 50090474-50090474 on Build GRCh38
rs528372280 Chr.22: 50090518-50090518 on Build GRCh38
rs767463723 Chr.22: 50090449-50090449 on Build GRCh38
rs540267845 Chr.22: 50090448-50090448 on Build GRCh38
rs969949343 Chr.22: 50090537-50090537 on Build GRCh38
rs957153061 Chr.22: 50090549-50090549 on Build GRCh38
rs9617018 Chr.22: 50090487-50090487 on Build GRCh38
rs546325738 Chr.22: 50090540-50090540 on Build GRCh38
rs370165934 Chr.22: 50090445-50090445 on Build GRCh38
rs781740427 Chr.22: 50090559-50090559 on Build GRCh38
rs1017407954 Chr.22: 50090561-50090561 on Build GRCh38
rs771736749 Chr.22: 50090541-50090542 on Build GRCh38
rs184330045 Chr.22: 50090451-50090451 on Build GRCh38

View Full Product Details