Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100576621-100576866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRCH4
Entrez Gene ID
4034
Gene Name
leucine rich repeats and calponin homology domain containing 4
Gene Aliases
LRN, LRRN1, LRRN4, PP14183
UniGene
Hs.125742
Gene Chromosome Location
Chr.7: 100574011-100586188 on Build GRCh38
Transcripts
NM_001289934.1 NM_002319.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6702798 c.1551A>G p.S517S Chr.7: 100576695-100576695 on Build GRCh38
6587293 c.1469-6C>A p.? Chr.7: 100576783-100576783 on Build GRCh38
7272054 c.1469C>T p.A490V Chr.7: 100576777-100576777 on Build GRCh38
5510238 c.1487G>A p.R496Q Chr.7: 100576759-100576759 on Build GRCh38
6175561 c.1543A>G p.S515G Chr.7: 100576703-100576703 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs937359122 Chr.7: 100576751-100576751 on Build GRCh38
rs774801531 Chr.7: 100576738-100576738 on Build GRCh38
rs778267206 Chr.7: 100576684-100576684 on Build GRCh38
rs778684939 Chr.7: 100576644-100576644 on Build GRCh38
rs372621081 Chr.7: 100576776-100576776 on Build GRCh38
rs765077735 Chr.7: 100576768-100576768 on Build GRCh38
rs185795340 Chr.7: 100576786-100576786 on Build GRCh38
rs952165220 Chr.7: 100576756-100576756 on Build GRCh38
rs756521295 Chr.7: 100576679-100576679 on Build GRCh38
rs765920224 Chr.7: 100576674-100576688 on Build GRCh38
rs776533200 Chr.7: 100576747-100576747 on Build GRCh38
rs1054828804 Chr.7: 100576765-100576765 on Build GRCh38
rs767457229 Chr.7: 100576661-100576661 on Build GRCh38
rs376096274 Chr.7: 100576674-100576674 on Build GRCh38
rs761681238 Chr.7: 100576760-100576760 on Build GRCh38
rs779180341 Chr.7: 100576843-100576843 on Build GRCh38
rs867804306 Chr.7: 100576694-100576694 on Build GRCh38
rs754423247 Chr.7: 100576830-100576831 on Build GRCh38
rs780574244 Chr.7: 100576803-100576803 on Build GRCh38
rs902332677 Chr.7: 100576816-100576816 on Build GRCh38
rs532308510 Chr.7: 100576643-100576643 on Build GRCh38
rs750086082 Chr.7: 100576846-100576846 on Build GRCh38
rs745480050 Chr.7: 100576718-100576718 on Build GRCh38
rs756950523 Chr.7: 100576713-100576713 on Build GRCh38
rs1042266197 Chr.7: 100576813-100576813 on Build GRCh38
rs370077970 Chr.7: 100576757-100576757 on Build GRCh38
rs919425885 Chr.7: 100576755-100576755 on Build GRCh38
rs752714318 Chr.7: 100576785-100576785 on Build GRCh38
rs76551586 Chr.7: 100576769-100576769 on Build GRCh38
rs747021778 Chr.7: 100576740-100576740 on Build GRCh38
rs749584458 Chr.7: 100576701-100576701 on Build GRCh38
rs368786964 Chr.7: 100576835-100576835 on Build GRCh38
rs1040078442 Chr.7: 100576799-100576799 on Build GRCh38
rs991531339 Chr.7: 100576682-100576682 on Build GRCh38
rs778396937 Chr.7: 100576833-100576834 on Build GRCh38
rs150672994 Chr.7: 100576717-100576717 on Build GRCh38
rs376880398 Chr.7: 100576794-100576794 on Build GRCh38
rs745582986 Chr.7: 100576750-100576750 on Build GRCh38
rs997992782 Chr.7: 100576840-100576840 on Build GRCh38
rs371513886 Chr.7: 100576807-100576807 on Build GRCh38
rs768477550 Chr.7: 100576744-100576744 on Build GRCh38
rs922896695 Chr.7: 100576800-100576800 on Build GRCh38
rs777525615 Chr.7: 100576673-100576673 on Build GRCh38
rs751559503 Chr.7: 100576737-100576737 on Build GRCh38
rs755132798 Chr.7: 100576648-100576648 on Build GRCh38
rs767650564 Chr.7: 100576777-100576777 on Build GRCh38
rs753506857 Chr.7: 100576806-100576807 on Build GRCh38
rs972522356 Chr.7: 100576745-100576745 on Build GRCh38
rs757217316 Chr.7: 100576802-100576802 on Build GRCh38

View Full Product Details