Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 4188758-4189005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTOP1
Entrez Gene ID
133060
Gene Name
otopetrin 1
Gene Aliases
-
UniGene
Hs.534544
Gene Chromosome Location
Chr.4: 4188803-4226894 on Build GRCh38
Transcripts
NM_177998.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1618832 c.1669-2_1669delAGC p.? Chr.4: 4188973-4188975 on Build GRCh38
359880 c.1671T>C p.L557L Chr.4: 4188971-4188971 on Build GRCh38
4005768 c.1792C>G p.R598G Chr.4: 4188850-4188850 on Build GRCh38
3603900 c.1749G>A p.W583* Chr.4: 4188893-4188893 on Build GRCh38
5034035 c.1681C>G p.P561A Chr.4: 4188961-4188961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200302629 Chr.4: 4188968-4188968 on Build GRCh38
rs759309550 Chr.4: 4188867-4188867 on Build GRCh38
rs749640790 Chr.4: 4188967-4188967 on Build GRCh38
rs763386902 Chr.4: 4188784-4188785 on Build GRCh38
rs201195209 Chr.4: 4188798-4188798 on Build GRCh38
rs746228046 Chr.4: 4188841-4188841 on Build GRCh38
rs764106951 Chr.4: 4188958-4188958 on Build GRCh38
rs141563893 Chr.4: 4188938-4188938 on Build GRCh38
rs868294160 Chr.4: 4188838-4188838 on Build GRCh38
rs753446220 Chr.4: 4188852-4188852 on Build GRCh38
rs764993571 Chr.4: 4188950-4188950 on Build GRCh38
rs188805040 Chr.4: 4188809-4188809 on Build GRCh38
rs776308014 Chr.4: 4188942-4188942 on Build GRCh38
rs200368405 Chr.4: 4188850-4188850 on Build GRCh38
rs375899210 Chr.4: 4188842-4188842 on Build GRCh38
rs750609534 Chr.4: 4188964-4188964 on Build GRCh38
rs748040258 Chr.4: 4188803-4188803 on Build GRCh38
rs770430029 Chr.4: 4188977-4188977 on Build GRCh38
rs753665250 Chr.4: 4188797-4188797 on Build GRCh38
rs879585365 Chr.4: 4188877-4188877 on Build GRCh38
rs538042112 Chr.4: 4188898-4188898 on Build GRCh38
rs760122186 Chr.4: 4188934-4188934 on Build GRCh38
rs762911635 Chr.4: 4188956-4188956 on Build GRCh38
rs749700109 Chr.4: 4188917-4188917 on Build GRCh38
rs149188671 Chr.4: 4188859-4188859 on Build GRCh38
rs781161496 Chr.4: 4188961-4188961 on Build GRCh38
rs758517130 Chr.4: 4188804-4188804 on Build GRCh38
rs769118497 Chr.4: 4188919-4188919 on Build GRCh38
rs757334904 Chr.4: 4188960-4188960 on Build GRCh38
rs79218997 Chr.4: 4188868-4188868 on Build GRCh38
rs759129260 Chr.4: 4188981-4188981 on Build GRCh38
rs147866723 Chr.4: 4188888-4188888 on Build GRCh38
rs748685757 Chr.4: 4188976-4188976 on Build GRCh38
rs770156025 Chr.4: 4188915-4188915 on Build GRCh38
rs779325797 Chr.4: 4188974-4188974 on Build GRCh38
rs554408396 Chr.4: 4188945-4188945 on Build GRCh38
rs568991103 Chr.4: 4188892-4188892 on Build GRCh38
rs755785191 Chr.4: 4188784-4188784 on Build GRCh38
rs774898827 Chr.4: 4188931-4188931 on Build GRCh38
rs775985932 Chr.4: 4188980-4188980 on Build GRCh38
rs771250229 Chr.4: 4188827-4188827 on Build GRCh38
rs747137441 Chr.4: 4188820-4188820 on Build GRCh38
rs199847133 Chr.4: 4188799-4188799 on Build GRCh38
rs777900732 Chr.4: 4188893-4188893 on Build GRCh38
rs144725579 Chr.4: 4188822-4188822 on Build GRCh38
rs780304132 Chr.4: 4188916-4188916 on Build GRCh38
rs1041743761 Chr.4: 4188928-4188928 on Build GRCh38
rs147648770 Chr.4: 4188959-4188959 on Build GRCh38
rs746077988 Chr.4: 4188911-4188911 on Build GRCh38
rs781414142 Chr.4: 4188909-4188909 on Build GRCh38
rs377255503 Chr.4: 4188790-4188790 on Build GRCh38
rs199742451 Chr.4: 4188849-4188849 on Build GRCh38
rs776782100 Chr.4: 4188830-4188830 on Build GRCh38
rs757467521 Chr.4: 4188905-4188905 on Build GRCh38

View Full Product Details