Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 77570973-77571246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN12
Entrez Gene ID
5782
Gene Name
protein tyrosine phosphatase, non-receptor type 12
Gene Aliases
PTP-PEST, PTPG1
UniGene
Hs.61812
Gene Chromosome Location
Chr.7: 77537160-77640071 on Build GRCh38
Transcripts
NM_001131008.1 NM_001131009.1 NM_002835.3 XM_005250518.2 XM_006716073.3 XM_011516444.1 XM_017012474.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6222383 c.208+1G>T p.? Chr.7: 77571187-77571187 on Build GRCh38
236839 c.191A>G p.Y64C Chr.7: 77571169-77571169 on Build GRCh38
5019354 c.100-3A>T p.? Chr.7: 77571075-77571075 on Build GRCh38
5925386 c.145C>T p.P49S Chr.7: 77571123-77571123 on Build GRCh38
7308195 c.144T>C p.Y48Y Chr.7: 77571122-77571122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045315245 Chr.7: 77571118-77571118 on Build GRCh38
rs564768286 Chr.7: 77571130-77571130 on Build GRCh38
rs754145211 Chr.7: 77571155-77571155 on Build GRCh38
rs904102433 Chr.7: 77571152-77571152 on Build GRCh38
rs750529622 Chr.7: 77571070-77571071 on Build GRCh38
rs189188150 Chr.7: 77571035-77571035 on Build GRCh38
rs758491437 Chr.7: 77571214-77571214 on Build GRCh38
rs372528657 Chr.7: 77571083-77571083 on Build GRCh38
rs928753088 Chr.7: 77571038-77571038 on Build GRCh38
rs146138004 Chr.7: 77571043-77571043 on Build GRCh38
rs200867808 Chr.7: 77571078-77571078 on Build GRCh38
rs764526702 Chr.7: 77571137-77571137 on Build GRCh38
rs759081456 Chr.7: 77571126-77571126 on Build GRCh38
rs750543432 Chr.7: 77571200-77571200 on Build GRCh38
rs930012124 Chr.7: 77571032-77571032 on Build GRCh38
rs982810965 Chr.7: 77571038-77571039 on Build GRCh38
rs919897977 Chr.7: 77571008-77571008 on Build GRCh38
rs544040347 Chr.7: 77571007-77571007 on Build GRCh38
rs765694045 Chr.7: 77571185-77571185 on Build GRCh38
rs778979752 Chr.7: 77571065-77571065 on Build GRCh38
rs17155183 Chr.7: 77571075-77571075 on Build GRCh38
rs199565508 Chr.7: 77571122-77571122 on Build GRCh38
rs974303214 Chr.7: 77571000-77571000 on Build GRCh38
rs761014478 Chr.7: 77571133-77571133 on Build GRCh38
rs770164884 Chr.7: 77571164-77571164 on Build GRCh38
rs767845789 Chr.7: 77571056-77571056 on Build GRCh38
rs774025235 Chr.7: 77571125-77571125 on Build GRCh38
rs1052505118 Chr.7: 77571180-77571180 on Build GRCh38
rs749249029 Chr.7: 77571117-77571117 on Build GRCh38
rs747950250 Chr.7: 77571079-77571079 on Build GRCh38
rs769625705 Chr.7: 77571080-77571080 on Build GRCh38
rs764571792 Chr.7: 77571034-77571034 on Build GRCh38
rs121434623 Chr.7: 77571160-77571160 on Build GRCh38

View Full Product Details