Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2061059-2061302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAT8L
Entrez Gene ID
339983
Gene Name
N-acetyltransferase 8 like
Gene Aliases
CML3, NACED, NAT8-LIKE
UniGene
Hs.318529
Gene Chromosome Location
Chr.4: 2059512-2069089 on Build GRCh38
Transcripts
NM_178557.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5718912 c.496C>A p.H166N Chr.4: 2061117-2061117 on Build GRCh38
1733815 c.32C>T p.P11L Chr.4: 2061157-2061157 on Build GRCh38
1733814 c.536C>T p.P179L Chr.4: 2061157-2061157 on Build GRCh38
6269282 c.483G>T p.L161L Chr.4: 2061104-2061104 on Build GRCh38
6722044 c.515T>C p.I172T Chr.4: 2061136-2061136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774180725 Chr.4: 2061130-2061130 on Build GRCh38
rs978794279 Chr.4: 2061260-2061260 on Build GRCh38
rs753331710 Chr.4: 2061167-2061167 on Build GRCh38
rs750896978 Chr.4: 2061086-2061086 on Build GRCh38
rs911223793 Chr.4: 2061261-2061261 on Build GRCh38
rs185577409 Chr.4: 2061218-2061218 on Build GRCh38
rs968986793 Chr.4: 2061248-2061248 on Build GRCh38
rs755328893 Chr.4: 2061174-2061174 on Build GRCh38
rs781706899 Chr.4: 2061177-2061177 on Build GRCh38
rs1017467018 Chr.4: 2061106-2061106 on Build GRCh38
rs752926170 Chr.4: 2061178-2061178 on Build GRCh38
rs781630013 Chr.4: 2061104-2061104 on Build GRCh38
rs752021014 Chr.4: 2061094-2061094 on Build GRCh38
rs559522261 Chr.4: 2061188-2061188 on Build GRCh38
rs976841486 Chr.4: 2061272-2061272 on Build GRCh38
rs755865891 Chr.4: 2061112-2061112 on Build GRCh38
rs942754156 Chr.4: 2061155-2061155 on Build GRCh38
rs1003242925 Chr.4: 2061211-2061211 on Build GRCh38
rs776193690 Chr.4: 2061203-2061203 on Build GRCh38
rs943102120 Chr.4: 2061280-2061280 on Build GRCh38
rs759388593 Chr.4: 2061131-2061131 on Build GRCh38
rs901718520 Chr.4: 2061126-2061126 on Build GRCh38
rs969141930 Chr.4: 2061208-2061208 on Build GRCh38
rs778752639 Chr.4: 2061190-2061190 on Build GRCh38
rs755238942 Chr.4: 2061102-2061102 on Build GRCh38
rs748394253 Chr.4: 2061107-2061107 on Build GRCh38
rs775068714 Chr.4: 2061159-2061159 on Build GRCh38
rs764809085 Chr.4: 2061207-2061207 on Build GRCh38
rs565270755 Chr.4: 2061200-2061200 on Build GRCh38
rs775297838 Chr.4: 2061198-2061198 on Build GRCh38
rs1038852207 Chr.4: 2061283-2061283 on Build GRCh38
rs200888256 Chr.4: 2061170-2061170 on Build GRCh38
rs1047434435 Chr.4: 2061183-2061183 on Build GRCh38
rs199612782 Chr.4: 2061160-2061160 on Build GRCh38
rs190005075 Chr.4: 2061270-2061270 on Build GRCh38
rs769862104 Chr.4: 2061113-2061113 on Build GRCh38
rs1048863812 Chr.4: 2061081-2061081 on Build GRCh38
rs543109424 Chr.4: 2061089-2061089 on Build GRCh38
rs1030068114 Chr.4: 2061158-2061158 on Build GRCh38
rs114826185 Chr.4: 2061194-2061194 on Build GRCh38
rs761296574 Chr.4: 2061206-2061206 on Build GRCh38
rs887594445 Chr.4: 2061085-2061085 on Build GRCh38
rs960490507 Chr.4: 2061212-2061212 on Build GRCh38
rs377569368 Chr.4: 2061180-2061180 on Build GRCh38
rs369632714 Chr.4: 2061169-2061169 on Build GRCh38
rs751967889 Chr.4: 2061171-2061171 on Build GRCh38
rs143186812 Chr.4: 2061141-2061141 on Build GRCh38
rs760151791 Chr.4: 2061211-2061211 on Build GRCh38
rs369480209 Chr.4: 2061093-2061093 on Build GRCh38
rs768242779 Chr.4: 2061202-2061202 on Build GRCh38
rs774949352 Chr.4: 2061210-2061210 on Build GRCh38
rs749383870 Chr.4: 2061184-2061184 on Build GRCh38
rs370820367 Chr.4: 2061122-2061122 on Build GRCh38
rs763623026 Chr.4: 2061162-2061162 on Build GRCh38
rs562996664 Chr.4: 2061154-2061154 on Build GRCh38
rs767961381 Chr.4: 2061214-2061214 on Build GRCh38
rs997450327 Chr.4: 2061150-2061150 on Build GRCh38
rs376253416 Chr.4: 2061161-2061161 on Build GRCh38
rs758862869 Chr.4: 2061092-2061092 on Build GRCh38
rs193116523 Chr.4: 2061195-2061195 on Build GRCh38

View Full Product Details