Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60946639-60946912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC15A3
Entrez Gene ID
51296
Gene Name
solute carrier family 15 member 3
Gene Aliases
OCTP, PHT2, PTR3
UniGene
Hs.237856
Gene Chromosome Location
Chr.11: 60937083-60952081 on Build GRCh38
Transcripts
NM_016582.2 NR_027391.1 XM_011545095.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1127717 c.560T>G p.V187G Chr.11: 60946820-60946820 on Build GRCh38
5453954 c.627T>C p.G209G Chr.11: 60946753-60946753 on Build GRCh38
6779727 c.711G>A p.V237V Chr.11: 60946669-60946669 on Build GRCh38
1225938 c.604T>G p.F202V Chr.11: 60946776-60946776 on Build GRCh38
467078 c.682C>A p.L228M Chr.11: 60946698-60946698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576463224 Chr.11: 60946765-60946765 on Build GRCh38
rs528183979 Chr.11: 60946791-60946791 on Build GRCh38
rs114034576 Chr.11: 60946804-60946804 on Build GRCh38
rs778400478 Chr.11: 60946721-60946721 on Build GRCh38
rs375591770 Chr.11: 60946718-60946718 on Build GRCh38
rs371715179 Chr.11: 60946844-60946844 on Build GRCh38
rs544936334 Chr.11: 60946710-60946710 on Build GRCh38
rs899259928 Chr.11: 60946685-60946685 on Build GRCh38
rs780323606 Chr.11: 60946839-60946839 on Build GRCh38
rs748144619 Chr.11: 60946859-60946859 on Build GRCh38
rs747174620 Chr.11: 60946843-60946843 on Build GRCh38
rs749740020 Chr.11: 60946793-60946793 on Build GRCh38
rs932284416 Chr.11: 60946885-60946885 on Build GRCh38
rs940658200 Chr.11: 60946869-60946869 on Build GRCh38
rs767081932 Chr.11: 60946810-60946810 on Build GRCh38
rs748692683 Chr.11: 60946716-60946716 on Build GRCh38
rs367882663 Chr.11: 60946764-60946764 on Build GRCh38
rs1037724779 Chr.11: 60946871-60946871 on Build GRCh38
rs757578910 Chr.11: 60946794-60946794 on Build GRCh38
rs750065131 Chr.11: 60946776-60946776 on Build GRCh38
rs890618349 Chr.11: 60946768-60946768 on Build GRCh38
rs746293686 Chr.11: 60946825-60946825 on Build GRCh38
rs751160363 Chr.11: 60946787-60946787 on Build GRCh38
rs764069229 Chr.11: 60946760-60946760 on Build GRCh38
rs772321643 Chr.11: 60946800-60946800 on Build GRCh38
rs900768268 Chr.11: 60946754-60946754 on Build GRCh38
rs773095907 Chr.11: 60946806-60946806 on Build GRCh38
rs541552550 Chr.11: 60946795-60946795 on Build GRCh38
rs757747133 Chr.11: 60946819-60946819 on Build GRCh38
rs754328027 Chr.11: 60946812-60946812 on Build GRCh38
rs746111304 Chr.11: 60946797-60946797 on Build GRCh38
rs768719882 Chr.11: 60946802-60946802 on Build GRCh38
rs778489653 Chr.11: 60946705-60946705 on Build GRCh38
rs377073448 Chr.11: 60946742-60946742 on Build GRCh38
rs775508489 Chr.11: 60946757-60946757 on Build GRCh38
rs750071947 Chr.11: 60946839-60946840 on Build GRCh38
rs147928255 Chr.11: 60946731-60946731 on Build GRCh38
rs771325349 Chr.11: 60946727-60946727 on Build GRCh38
rs369804608 Chr.11: 60946828-60946828 on Build GRCh38
rs765024044 Chr.11: 60946767-60946767 on Build GRCh38
rs142952480 Chr.11: 60946790-60946790 on Build GRCh38
rs773005748 Chr.11: 60946866-60946866 on Build GRCh38
rs140918009 Chr.11: 60946809-60946809 on Build GRCh38
rs201623464 Chr.11: 60946820-60946820 on Build GRCh38
rs749670306 Chr.11: 60946726-60946726 on Build GRCh38
rs781291333 Chr.11: 60946851-60946851 on Build GRCh38
rs1052452661 Chr.11: 60946888-60946888 on Build GRCh38
rs776444965 Chr.11: 60946803-60946803 on Build GRCh38
rs747151856 Chr.11: 60946801-60946801 on Build GRCh38
rs545409760 Chr.11: 60946784-60946784 on Build GRCh38
rs1030958495 Chr.11: 60946772-60946772 on Build GRCh38
rs149019885 Chr.11: 60946741-60946741 on Build GRCh38
rs767214475 Chr.11: 60946787-60946787 on Build GRCh38
rs762816229 Chr.11: 60946807-60946807 on Build GRCh38
rs769814913 Chr.11: 60946864-60946864 on Build GRCh38
rs1039756370 Chr.11: 60946744-60946744 on Build GRCh38
rs764972944 Chr.11: 60946805-60946805 on Build GRCh38
rs758029103 Chr.11: 60946778-60946778 on Build GRCh38
rs372297850 Chr.11: 60946666-60946666 on Build GRCh38
rs750023687 Chr.11: 60946706-60946706 on Build GRCh38
rs760715158 Chr.11: 60946759-60946759 on Build GRCh38
rs758750713 Chr.11: 60946832-60946832 on Build GRCh38

View Full Product Details