Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 127886558-127886831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLK4
Entrez Gene ID
10733
Gene Name
polo like kinase 4
Gene Aliases
MCCRP2, SAK, STK18
UniGene
Hs.172052
Gene Chromosome Location
Chr.4: 127880861-127899224 on Build GRCh38
Transcripts
NM_001190799.1 NM_001190801.1 NM_014264.4 XM_005262701.2 XM_017007662.1 XM_017007663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5727624 c.1310C>T p.T437I Chr.4: 127886680-127886680 on Build GRCh38
1051121 c.1322C>A p.S441Y Chr.4: 127886692-127886692 on Build GRCh38
7166596 c.1252G>C p.E418Q Chr.4: 127886622-127886622 on Build GRCh38
5523519 c.1303G>A p.E435K Chr.4: 127886673-127886673 on Build GRCh38
3600195 c.1340C>T p.P447L Chr.4: 127886710-127886710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754107191 Chr.4: 127886650-127886650 on Build GRCh38
rs781121267 Chr.4: 127886717-127886720 on Build GRCh38
rs144633592 Chr.4: 127886589-127886589 on Build GRCh38
rs34906574 Chr.4: 127886715-127886715 on Build GRCh38
rs781315361 Chr.4: 127886688-127886688 on Build GRCh38
rs773353771 Chr.4: 127886599-127886599 on Build GRCh38
rs769597103 Chr.4: 127886596-127886596 on Build GRCh38
rs114505904 Chr.4: 127886793-127886793 on Build GRCh38
rs755956447 Chr.4: 127886714-127886714 on Build GRCh38
rs142775450 Chr.4: 127886713-127886713 on Build GRCh38
rs777061496 Chr.4: 127886748-127886748 on Build GRCh38
rs748018322 Chr.4: 127886591-127886591 on Build GRCh38
rs1010560546 Chr.4: 127886729-127886729 on Build GRCh38
rs769006836 Chr.4: 127886743-127886743 on Build GRCh38
rs746015384 Chr.4: 127886728-127886728 on Build GRCh38
rs771016420 Chr.4: 127886621-127886621 on Build GRCh38
rs762067355 Chr.4: 127886771-127886771 on Build GRCh38
rs376946475 Chr.4: 127886611-127886611 on Build GRCh38
rs761271378 Chr.4: 127886742-127886742 on Build GRCh38
rs199630762 Chr.4: 127886673-127886673 on Build GRCh38
rs772109280 Chr.4: 127886733-127886733 on Build GRCh38
rs724159996 Chr.4: 127886669-127886673 on Build GRCh38
rs530079860 Chr.4: 127886805-127886805 on Build GRCh38
rs369153451 Chr.4: 127886624-127886624 on Build GRCh38
rs111593796 Chr.4: 127886644-127886644 on Build GRCh38
rs775972407 Chr.4: 127886629-127886629 on Build GRCh38
rs768073163 Chr.4: 127886627-127886627 on Build GRCh38
rs563501150 Chr.4: 127886719-127886719 on Build GRCh38
rs751170471 Chr.4: 127886682-127886682 on Build GRCh38
rs945476728 Chr.4: 127886770-127886772 on Build GRCh38
rs751221778 Chr.4: 127886776-127886776 on Build GRCh38
rs943317577 Chr.4: 127886623-127886623 on Build GRCh38
rs766199080 Chr.4: 127886774-127886774 on Build GRCh38
rs903888281 Chr.4: 127886804-127886804 on Build GRCh38
rs766069077 Chr.4: 127886680-127886680 on Build GRCh38
rs764500240 Chr.4: 127886640-127886640 on Build GRCh38
rs774545143 Chr.4: 127886726-127886726 on Build GRCh38
rs138185535 Chr.4: 127886622-127886622 on Build GRCh38
rs771156770 Chr.4: 127886720-127886720 on Build GRCh38
rs1056959225 Chr.4: 127886703-127886703 on Build GRCh38
rs776029663 Chr.4: 127886738-127886738 on Build GRCh38

View Full Product Details