Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10849279-10849526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRH1-PRR4
Entrez Gene ID
100533464
Gene Name
PRH1-PRR4 readthrough
Gene Aliases
PRH1
UniGene
Hs.732031
Gene Chromosome Location
Chr.12: 10845849-11171625 on Build GRCh38
Transcripts
NR_037918.2 NM_001098538.2 NM_007244.2

Gene Symbol
PRR4
Entrez Gene ID
11272
Gene Name
proline rich 4 (lacrimal)
Gene Aliases
LPRP, PROL4
UniGene
Hs.732031
Gene Chromosome Location
Chr.12: 10845849-10849476 on Build GRCh38
Transcripts
NR_037918.2 NM_001098538.2 NM_007244.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
430302 c.49C>G p.Q17E Chr.12: 10849389-10849389 on Build GRCh38
1946621 c.20C>T p.S7L Chr.12: 10849418-10849418 on Build GRCh38
5766812 c.64+10T>A p.? Chr.12: 10849364-10849364 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769243923 Chr.12: 10849352-10849352 on Build GRCh38
rs766526937 Chr.12: 10849391-10849392 on Build GRCh38
rs747516829 Chr.12: 10849437-10849437 on Build GRCh38
rs1029664318 Chr.12: 10849492-10849492 on Build GRCh38
rs544091066 Chr.12: 10849348-10849348 on Build GRCh38
rs977082585 Chr.12: 10849429-10849429 on Build GRCh38
rs766299863 Chr.12: 10849473-10849473 on Build GRCh38
rs761870266 Chr.12: 10849354-10849355 on Build GRCh38
rs776354046 Chr.12: 10849348-10849349 on Build GRCh38
rs758827787 Chr.12: 10849432-10849432 on Build GRCh38
rs137863298 Chr.12: 10849367-10849367 on Build GRCh38
rs774084101 Chr.12: 10849455-10849455 on Build GRCh38
rs200572152 Chr.12: 10849364-10849364 on Build GRCh38
rs1025720229 Chr.12: 10849350-10849350 on Build GRCh38
rs185779358 Chr.12: 10849384-10849384 on Build GRCh38
rs905598849 Chr.12: 10849309-10849309 on Build GRCh38
rs752972789 Chr.12: 10849489-10849489 on Build GRCh38
rs778096426 Chr.12: 10849434-10849434 on Build GRCh38
rs762549589 Chr.12: 10849365-10849365 on Build GRCh38
rs895771611 Chr.12: 10849339-10849339 on Build GRCh38
rs201610973 Chr.12: 10849365-10849365 on Build GRCh38
rs753062999 Chr.12: 10849425-10849425 on Build GRCh38
rs779457651 Chr.12: 10849440-10849440 on Build GRCh38
rs774313938 Chr.12: 10849372-10849372 on Build GRCh38
rs188420777 Chr.12: 10849299-10849299 on Build GRCh38
rs748462894 Chr.12: 10849335-10849335 on Build GRCh38
rs201802308 Chr.12: 10849472-10849472 on Build GRCh38
rs748779044 Chr.12: 10849447-10849447 on Build GRCh38
rs375138635 Chr.12: 10849439-10849439 on Build GRCh38
rs766419384 Chr.12: 10849388-10849388 on Build GRCh38
rs375673181 Chr.12: 10849469-10849469 on Build GRCh38
rs750564315 Chr.12: 10849364-10849365 on Build GRCh38
rs762964216 Chr.12: 10849386-10849386 on Build GRCh38
rs1024227643 Chr.12: 10849300-10849300 on Build GRCh38
rs747544643 Chr.12: 10849349-10849349 on Build GRCh38
rs768445299 Chr.12: 10849337-10849337 on Build GRCh38
rs759544041 Chr.12: 10849484-10849484 on Build GRCh38
rs750454758 Chr.12: 10849383-10849383 on Build GRCh38
rs761646506 Chr.12: 10849375-10849375 on Build GRCh38
rs779049143 Chr.12: 10849420-10849420 on Build GRCh38
rs534160978 Chr.12: 10849361-10849361 on Build GRCh38
rs753923856 Chr.12: 10849409-10849409 on Build GRCh38
rs753877148 Chr.12: 10849477-10849477 on Build GRCh38
rs555027683 Chr.12: 10849474-10849474 on Build GRCh38
rs767570416 Chr.12: 10849378-10849378 on Build GRCh38
rs956657671 Chr.12: 10849496-10849496 on Build GRCh38
rs768439991 Chr.12: 10849454-10849454 on Build GRCh38
rs761554843 Chr.12: 10849463-10849463 on Build GRCh38
rs755187303 Chr.12: 10849410-10849410 on Build GRCh38
rs775040339 Chr.12: 10849355-10849355 on Build GRCh38
rs1046051585 Chr.12: 10849504-10849504 on Build GRCh38
rs180884675 Chr.12: 10849323-10849323 on Build GRCh38
rs994051191 Chr.12: 10849357-10849357 on Build GRCh38
rs765295108 Chr.12: 10849488-10849488 on Build GRCh38

View Full Product Details