Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137199715-137199970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPRN
Entrez Gene ID
286262
Gene Name
taperin
Gene Aliases
C9orf75, DFNB79
UniGene
Hs.323445
Gene Chromosome Location
Chr.9: 137191617-137200711 on Build GRCh38
Transcripts
NM_001128228.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4514605 c.62C>T p.S21F Chr.9: 137199732-137199732 on Build GRCh38
4389458 c.779A>C p.H260P Chr.9: 137199933-137199933 on Build GRCh38
6406701 c.985A>G p.N329D Chr.9: 137199727-137199727 on Build GRCh38
4514845 c.71C>T p.S24F Chr.9: 137199723-137199723 on Build GRCh38
5153371 c.56C>G p.A19G Chr.9: 137199738-137199738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992879712 Chr.9: 137199823-137199823 on Build GRCh38
rs535676505 Chr.9: 137199848-137199848 on Build GRCh38
rs200016367 Chr.9: 137199742-137199742 on Build GRCh38
rs147404722 Chr.9: 137199821-137199821 on Build GRCh38
rs771154279 Chr.9: 137199815-137199815 on Build GRCh38
rs745997859 Chr.9: 137199942-137199942 on Build GRCh38
rs754757066 Chr.9: 137199868-137199868 on Build GRCh38
rs767394503 Chr.9: 137199778-137199778 on Build GRCh38
rs751049185 Chr.9: 137199767-137199767 on Build GRCh38
rs749999995 Chr.9: 137199849-137199849 on Build GRCh38
rs750372412 Chr.9: 137199779-137199779 on Build GRCh38
rs758757835 Chr.9: 137199870-137199896 on Build GRCh38
rs753049274 Chr.9: 137199818-137199819 on Build GRCh38
rs923699112 Chr.9: 137199943-137199943 on Build GRCh38
rs375619082 Chr.9: 137199854-137199854 on Build GRCh38
rs539737661 Chr.9: 137199818-137199818 on Build GRCh38
rs1000547607 Chr.9: 137199809-137199809 on Build GRCh38
rs762658637 Chr.9: 137199761-137199761 on Build GRCh38
rs781411318 Chr.9: 137199927-137199927 on Build GRCh38
rs1039638720 Chr.9: 137199785-137199785 on Build GRCh38
rs867073404 Chr.9: 137199933-137199933 on Build GRCh38
rs765704758 Chr.9: 137199755-137199755 on Build GRCh38
rs184862728 Chr.9: 137199871-137199871 on Build GRCh38
rs145558759 Chr.9: 137199743-137199743 on Build GRCh38
rs867531473 Chr.9: 137199936-137199936 on Build GRCh38
rs368102306 Chr.9: 137199762-137199762 on Build GRCh38
rs761256786 Chr.9: 137199841-137199841 on Build GRCh38
rs773327512 Chr.9: 137199740-137199740 on Build GRCh38
rs756793197 Chr.9: 137199769-137199769 on Build GRCh38
rs755838477 Chr.9: 137199780-137199780 on Build GRCh38
rs747130999 Chr.9: 137199741-137199741 on Build GRCh38
rs576209587 Chr.9: 137199830-137199830 on Build GRCh38
rs779894755 Chr.9: 137199781-137199781 on Build GRCh38
rs972076137 Chr.9: 137199858-137199858 on Build GRCh38
rs557115232 Chr.9: 137199861-137199861 on Build GRCh38
rs374735561 Chr.9: 137199760-137199760 on Build GRCh38
rs763762194 Chr.9: 137199827-137199827 on Build GRCh38
rs757623362 Chr.9: 137199880-137199880 on Build GRCh38
rs904429045 Chr.9: 137199808-137199808 on Build GRCh38
rs749094012 Chr.9: 137199793-137199793 on Build GRCh38
rs758620993 Chr.9: 137199802-137199802 on Build GRCh38
rs148880118 Chr.9: 137199747-137199747 on Build GRCh38
rs971540821 Chr.9: 137199928-137199928 on Build GRCh38
rs751841555 Chr.9: 137199873-137199873 on Build GRCh38
rs765756665 Chr.9: 137199768-137199769 on Build GRCh38

View Full Product Details