Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 7259358-7259591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STS
Entrez Gene ID
412
Gene Name
steroid sulfatase (microsomal), isozyme S
Gene Aliases
ARSC, ARSC1, ASC, ES, SSDD, XLI
UniGene
Hs.522578
Gene Chromosome Location
Chr.X: 7147252-7354643 on Build GRCh38
Transcripts
NM_000351.5 NM_001320750.1 NM_001320751.1 NM_001320752.1 NM_001320753.1 NM_001320754.1

SNP Detail

SNP ID Chromosome Location
rs148296838 Chr.X: 7259554-7259554 on Build GRCh38
rs1032020820 Chr.X: 7259490-7259490 on Build GRCh38
rs747592165 Chr.X: 7259385-7259385 on Build GRCh38
rs748874334 Chr.X: 7259530-7259530 on Build GRCh38
rs777387752 Chr.X: 7259528-7259528 on Build GRCh38
rs751989009 Chr.X: 7259494-7259494 on Build GRCh38
rs772693579 Chr.X: 7259406-7259406 on Build GRCh38
rs143711517 Chr.X: 7259421-7259421 on Build GRCh38
rs758308023 Chr.X: 7259503-7259503 on Build GRCh38
rs778962499 Chr.X: 7259472-7259472 on Build GRCh38
rs375875672 Chr.X: 7259413-7259413 on Build GRCh38
rs760966135 Chr.X: 7259401-7259401 on Build GRCh38
rs201808898 Chr.X: 7259414-7259414 on Build GRCh38
rs755543529 Chr.X: 7259527-7259527 on Build GRCh38
rs745598872 Chr.X: 7259569-7259569 on Build GRCh38
rs915654648 Chr.X: 7259512-7259512 on Build GRCh38
rs759538143 Chr.X: 7259430-7259430 on Build GRCh38
rs144195721 Chr.X: 7259476-7259476 on Build GRCh38
rs866913063 Chr.X: 7259543-7259543 on Build GRCh38
rs368690119 Chr.X: 7259469-7259469 on Build GRCh38
rs61733086 Chr.X: 7259507-7259507 on Build GRCh38
rs199596711 Chr.X: 7259402-7259402 on Build GRCh38
rs765388132 Chr.X: 7259467-7259467 on Build GRCh38
rs372283033 Chr.X: 7259491-7259491 on Build GRCh38
rs767492847 Chr.X: 7259445-7259445 on Build GRCh38
rs775579490 Chr.X: 7259464-7259464 on Build GRCh38
rs780677288 Chr.X: 7259377-7259377 on Build GRCh38
rs369481705 Chr.X: 7259531-7259531 on Build GRCh38
rs376588996 Chr.X: 7259518-7259518 on Build GRCh38
rs761932927 Chr.X: 7259465-7259465 on Build GRCh38

View Full Product Details