Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35744128-35744379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
U2AF1L4
Entrez Gene ID
199746
Gene Name
U2 small nuclear RNA auxiliary factor 1 like 4
Gene Aliases
U2AF1-RS3, U2AF1L3, U2AF1L3V1, U2AF1RS3, U2af26
UniGene
Hs.351558
Gene Chromosome Location
Chr.19: 35742527-35745442 on Build GRCh38
Transcripts
NM_001040425.2 NM_144987.3 NR_110171.1 NR_110172.1 NR_110173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
187988 c.189C>T p.C63C Chr.19: 35744365-35744365 on Build GRCh38
5892408 c.232-7C>T p.? Chr.19: 35744152-35744152 on Build GRCh38
7122183 c.177G>C p.E59D Chr.19: 35744377-35744377 on Build GRCh38
4902190 c.352C>T p.R118W Chr.19: 35744142-35744142 on Build GRCh38
7301683 c.220G>A p.V74I Chr.19: 35744334-35744334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752097505 Chr.19: 35744330-35744330 on Build GRCh38
rs755693473 Chr.19: 35744274-35744274 on Build GRCh38
rs1052111991 Chr.19: 35744164-35744164 on Build GRCh38
rs757670106 Chr.19: 35744173-35744175 on Build GRCh38
rs764561870 Chr.19: 35744327-35744327 on Build GRCh38
rs374231931 Chr.19: 35744273-35744273 on Build GRCh38
rs774313160 Chr.19: 35744307-35744307 on Build GRCh38
rs17851875 Chr.19: 35744346-35744346 on Build GRCh38
rs370791126 Chr.19: 35744319-35744319 on Build GRCh38
rs779652566 Chr.19: 35744306-35744307 on Build GRCh38
rs371520231 Chr.19: 35744310-35744310 on Build GRCh38
rs1005034610 Chr.19: 35744258-35744258 on Build GRCh38
rs751913089 Chr.19: 35744176-35744176 on Build GRCh38
rs1016397445 Chr.19: 35744277-35744277 on Build GRCh38
rs763097228 Chr.19: 35744323-35744323 on Build GRCh38
rs757625907 Chr.19: 35744343-35744343 on Build GRCh38
rs764624757 Chr.19: 35744175-35744175 on Build GRCh38
rs1003120808 Chr.19: 35744197-35744197 on Build GRCh38
rs1036391734 Chr.19: 35744208-35744208 on Build GRCh38
rs768609622 Chr.19: 35744284-35744284 on Build GRCh38
rs765509678 Chr.19: 35744315-35744315 on Build GRCh38
rs765412841 Chr.19: 35744152-35744152 on Build GRCh38
rs548534051 Chr.19: 35744329-35744329 on Build GRCh38
rs759757820 Chr.19: 35744148-35744148 on Build GRCh38
rs750725195 Chr.19: 35744354-35744354 on Build GRCh38
rs147371997 Chr.19: 35744183-35744183 on Build GRCh38
rs954567621 Chr.19: 35744347-35744347 on Build GRCh38
rs771897521 Chr.19: 35744311-35744311 on Build GRCh38
rs886589336 Chr.19: 35744230-35744230 on Build GRCh38
rs569684224 Chr.19: 35744251-35744251 on Build GRCh38
rs752768915 Chr.19: 35744174-35744174 on Build GRCh38
rs796512777 Chr.19: 35744325-35744325 on Build GRCh38

View Full Product Details