Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 100555307-100555534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM45A
Entrez Gene ID
55076
Gene Name
transmembrane protein 45A
Gene Aliases
DERP7, DNAPTP4
UniGene
Hs.658956
Gene Chromosome Location
Chr.3: 100492619-100577453 on Build GRCh38
Transcripts
NM_018004.2 XM_005247569.2 XM_017006715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5018594 c.143G>A p.R48Q Chr.3: 100555354-100555354 on Build GRCh38
359820 c.99G>C p.K33N Chr.3: 100555310-100555310 on Build GRCh38
1035623 c.186A>T p.L62F Chr.3: 100555397-100555397 on Build GRCh38
1327152 c.107G>A p.R36Q Chr.3: 100555318-100555318 on Build GRCh38
7107101 c.125C>A p.S42Y Chr.3: 100555336-100555336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146251365 Chr.3: 100555374-100555374 on Build GRCh38
rs143708068 Chr.3: 100555469-100555469 on Build GRCh38
rs372085362 Chr.3: 100555448-100555448 on Build GRCh38
rs753312840 Chr.3: 100555337-100555337 on Build GRCh38
rs545042972 Chr.3: 100555424-100555424 on Build GRCh38
rs947880127 Chr.3: 100555492-100555492 on Build GRCh38
rs55838872 Chr.3: 100555451-100555451 on Build GRCh38
rs200288076 Chr.3: 100555332-100555332 on Build GRCh38
rs767884040 Chr.3: 100555459-100555459 on Build GRCh38
rs149968178 Chr.3: 100555380-100555380 on Build GRCh38
rs1046575116 Chr.3: 100555460-100555460 on Build GRCh38
rs925233249 Chr.3: 100555343-100555343 on Build GRCh38
rs1021817771 Chr.3: 100555353-100555353 on Build GRCh38
rs779274823 Chr.3: 100555370-100555370 on Build GRCh38
rs909752346 Chr.3: 100555470-100555470 on Build GRCh38
rs562649568 Chr.3: 100555357-100555357 on Build GRCh38
rs758560405 Chr.3: 100555414-100555428 on Build GRCh38
rs755640348 Chr.3: 100555331-100555331 on Build GRCh38
rs910971463 Chr.3: 100555388-100555388 on Build GRCh38
rs1010882415 Chr.3: 100555338-100555338 on Build GRCh38
rs755527902 Chr.3: 100555366-100555366 on Build GRCh38
rs879056763 Chr.3: 100555403-100555403 on Build GRCh38
rs768699999 Chr.3: 100555437-100555437 on Build GRCh38
rs371533437 Chr.3: 100555409-100555409 on Build GRCh38
rs778071377 Chr.3: 100555393-100555393 on Build GRCh38
rs780213135 Chr.3: 100555426-100555426 on Build GRCh38
rs376157438 Chr.3: 100555414-100555414 on Build GRCh38
rs747180406 Chr.3: 100555408-100555408 on Build GRCh38
rs367901634 Chr.3: 100555433-100555433 on Build GRCh38

View Full Product Details