Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95494599-95494836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC37
Entrez Gene ID
9652
Gene Name
tetratricopeptide repeat domain 37
Gene Aliases
KIAA0372, Ski3, THES
UniGene
Hs.482868
Gene Chromosome Location
Chr.5: 95463895-95555005 on Build GRCh38
Transcripts
NM_014639.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
400027 c.3691-1G>T p.? Chr.5: 95494794-95494794 on Build GRCh38
4952440 c.3711G>C p.A1237A Chr.5: 95494773-95494773 on Build GRCh38
3941424 c.3780G>T p.Q1260H Chr.5: 95494704-95494704 on Build GRCh38
4160062 c.3743C>A p.A1248E Chr.5: 95494741-95494741 on Build GRCh38
6280205 c.3711G>A p.A1237A Chr.5: 95494773-95494773 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759920504 Chr.5: 95494724-95494724 on Build GRCh38
rs780501630 Chr.5: 95494638-95494639 on Build GRCh38
rs369531356 Chr.5: 95494801-95494801 on Build GRCh38
rs571064888 Chr.5: 95494782-95494782 on Build GRCh38
rs750228539 Chr.5: 95494775-95494775 on Build GRCh38
rs757122657 Chr.5: 95494676-95494676 on Build GRCh38
rs547538643 Chr.5: 95494721-95494721 on Build GRCh38
rs755499973 Chr.5: 95494632-95494632 on Build GRCh38
rs548734259 Chr.5: 95494690-95494690 on Build GRCh38
rs777598541 Chr.5: 95494675-95494675 on Build GRCh38
rs755317882 Chr.5: 95494800-95494804 on Build GRCh38
rs111235310 Chr.5: 95494719-95494719 on Build GRCh38
rs924502432 Chr.5: 95494629-95494629 on Build GRCh38
rs777893721 Chr.5: 95494802-95494802 on Build GRCh38
rs373311620 Chr.5: 95494694-95494694 on Build GRCh38
rs751111454 Chr.5: 95494766-95494766 on Build GRCh38
rs745666025 Chr.5: 95494686-95494686 on Build GRCh38
rs747074482 Chr.5: 95494807-95494807 on Build GRCh38
rs905539180 Chr.5: 95494756-95494756 on Build GRCh38
rs772138826 Chr.5: 95494712-95494712 on Build GRCh38
rs751964406 Chr.5: 95494639-95494639 on Build GRCh38
rs765760646 Chr.5: 95494652-95494652 on Build GRCh38
rs528862822 Chr.5: 95494666-95494666 on Build GRCh38
rs757861034 Chr.5: 95494742-95494742 on Build GRCh38
rs763630489 Chr.5: 95494743-95494743 on Build GRCh38
rs949843863 Chr.5: 95494734-95494734 on Build GRCh38
rs557540994 Chr.5: 95494774-95494774 on Build GRCh38
rs934570747 Chr.5: 95494677-95494677 on Build GRCh38
rs147416417 Chr.5: 95494773-95494773 on Build GRCh38
rs781201322 Chr.5: 95494679-95494679 on Build GRCh38
rs773432276 Chr.5: 95494715-95494715 on Build GRCh38
rs752197366 Chr.5: 95494653-95494653 on Build GRCh38
rs140236218 Chr.5: 95494733-95494733 on Build GRCh38
rs922595575 Chr.5: 95494622-95494622 on Build GRCh38
rs765703776 Chr.5: 95494731-95494731 on Build GRCh38
rs749188285 Chr.5: 95494790-95494790 on Build GRCh38
rs376534902 Chr.5: 95494711-95494711 on Build GRCh38
rs769672042 Chr.5: 95494689-95494689 on Build GRCh38
rs1057014596 Chr.5: 95494696-95494696 on Build GRCh38
rs765753375 Chr.5: 95494757-95494757 on Build GRCh38

View Full Product Details