Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122383594-122383867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAD1
Entrez Gene ID
132612
Gene Name
adenosine deaminase domain containing 1
Gene Aliases
Tenr
UniGene
Hs.518957
Gene Chromosome Location
Chr.4: 122378966-122429815 on Build GRCh38
Transcripts
NM_001159285.1 NM_001159295.1 NM_139243.3 XM_005262741.2 XM_005262743.2 XM_005262744.2 XM_005262745.2 XM_011531603.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4598961 c.424G>A p.G142R Chr.4: 122383861-122383861 on Build GRCh38
4799379 c.367G>A p.V123I Chr.4: 122383804-122383804 on Build GRCh38
1181760 c.416A>G p.Y139C Chr.4: 122383853-122383853 on Build GRCh38
6593312 c.384T>C p.F128F Chr.4: 122383821-122383821 on Build GRCh38
7123795 c.363T>A p.G121G Chr.4: 122383800-122383800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs915821713 Chr.4: 122383633-122383633 on Build GRCh38
rs746643559 Chr.4: 122383834-122383834 on Build GRCh38
rs62321702 Chr.4: 122383737-122383737 on Build GRCh38
rs756642898 Chr.4: 122383790-122383790 on Build GRCh38
rs527800757 Chr.4: 122383658-122383658 on Build GRCh38
rs985506611 Chr.4: 122383652-122383652 on Build GRCh38
rs759735464 Chr.4: 122383769-122383769 on Build GRCh38
rs199880328 Chr.4: 122383625-122383635 on Build GRCh38
rs772824110 Chr.4: 122383755-122383755 on Build GRCh38
rs868632817 Chr.4: 122383657-122383657 on Build GRCh38
rs767678669 Chr.4: 122383773-122383773 on Build GRCh38
rs966693607 Chr.4: 122383815-122383815 on Build GRCh38
rs572057798 Chr.4: 122383659-122383659 on Build GRCh38
rs747169138 Chr.4: 122383775-122383776 on Build GRCh38
rs779631119 Chr.4: 122383833-122383833 on Build GRCh38
rs763237947 Chr.4: 122383758-122383758 on Build GRCh38
rs757906078 Chr.4: 122383651-122383651 on Build GRCh38
rs780632203 Chr.4: 122383839-122383839 on Build GRCh38
rs774615295 Chr.4: 122383767-122383767 on Build GRCh38
rs1018925021 Chr.4: 122383672-122383672 on Build GRCh38
rs1017818226 Chr.4: 122383795-122383795 on Build GRCh38
rs764536577 Chr.4: 122383791-122383791 on Build GRCh38
rs780101676 Chr.4: 122383774-122383777 on Build GRCh38
rs998925938 Chr.4: 122383730-122383730 on Build GRCh38
rs750685928 Chr.4: 122383705-122383705 on Build GRCh38
rs564045534 Chr.4: 122383719-122383719 on Build GRCh38
rs754513934 Chr.4: 122383837-122383837 on Build GRCh38
rs757638868 Chr.4: 122383812-122383812 on Build GRCh38
rs965697981 Chr.4: 122383678-122383678 on Build GRCh38
rs754086597 Chr.4: 122383794-122383794 on Build GRCh38
rs753292237 Chr.4: 122383789-122383789 on Build GRCh38
rs766574993 Chr.4: 122383759-122383759 on Build GRCh38

View Full Product Details