Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62575396-62575643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUT1
Entrez Gene ID
64852
Gene Name
terminal uridylyl transferase 1, U6 snRNA-specific
Gene Aliases
PAPD2, RBM21, STARPAP, TUTase, URLC6
UniGene
Hs.728983
Gene Chromosome Location
Chr.11: 62575045-62591637 on Build GRCh38
Transcripts
NM_022830.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6996014 c.2281C>T p.P761S Chr.11: 62575438-62575438 on Build GRCh38
3450995 c.2112C>A p.A704A Chr.11: 62575607-62575607 on Build GRCh38
4146001 c.2261C>A p.A754E Chr.11: 62575458-62575458 on Build GRCh38
4840125 c.2193T>G p.V731V Chr.11: 62575640-62575640 on Build GRCh38
4430913 c.2104G>C p.D702H Chr.11: 62575615-62575615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746225172 Chr.11: 62575487-62575487 on Build GRCh38
rs781287321 Chr.11: 62575544-62575544 on Build GRCh38
rs765172205 Chr.11: 62575588-62575588 on Build GRCh38
rs752674302 Chr.11: 62575521-62575521 on Build GRCh38
rs550071205 Chr.11: 62575615-62575615 on Build GRCh38
rs143949073 Chr.11: 62575519-62575519 on Build GRCh38
rs369681798 Chr.11: 62575458-62575458 on Build GRCh38
rs762335509 Chr.11: 62575446-62575446 on Build GRCh38
rs752723975 Chr.11: 62575525-62575525 on Build GRCh38
rs770280915 Chr.11: 62575491-62575491 on Build GRCh38
rs761270793 Chr.11: 62575502-62575502 on Build GRCh38
rs761045437 Chr.11: 62575446-62575447 on Build GRCh38
rs930909781 Chr.11: 62575474-62575474 on Build GRCh38
rs564567770 Chr.11: 62575443-62575443 on Build GRCh38
rs759346476 Chr.11: 62575587-62575587 on Build GRCh38
rs142880507 Chr.11: 62575612-62575612 on Build GRCh38
rs746191567 Chr.11: 62575548-62575548 on Build GRCh38
rs753961345 Chr.11: 62575506-62575507 on Build GRCh38
rs886449265 Chr.11: 62575485-62575485 on Build GRCh38
rs750102769 Chr.11: 62575416-62575416 on Build GRCh38
rs35639864 Chr.11: 62575585-62575585 on Build GRCh38
rs36004630 Chr.11: 62575618-62575618 on Build GRCh38
rs144668445 Chr.11: 62575579-62575579 on Build GRCh38
rs765227246 Chr.11: 62575523-62575523 on Build GRCh38
rs758312026 Chr.11: 62575527-62575528 on Build GRCh38
rs996678716 Chr.11: 62575528-62575528 on Build GRCh38
rs747284522 Chr.11: 62575469-62575469 on Build GRCh38
rs149360817 Chr.11: 62575574-62575574 on Build GRCh38
rs759482013 Chr.11: 62575510-62575510 on Build GRCh38
rs771190111 Chr.11: 62575473-62575473 on Build GRCh38
rs748191823 Chr.11: 62575447-62575447 on Build GRCh38
rs373180120 Chr.11: 62575475-62575475 on Build GRCh38
rs755831260 Chr.11: 62575420-62575420 on Build GRCh38
rs138487018 Chr.11: 62575517-62575517 on Build GRCh38
rs369165209 Chr.11: 62575497-62575497 on Build GRCh38
rs771555615 Chr.11: 62575573-62575573 on Build GRCh38
rs34783570 Chr.11: 62575481-62575481 on Build GRCh38
rs750989080 Chr.11: 62575566-62575566 on Build GRCh38
rs766244715 Chr.11: 62575425-62575425 on Build GRCh38
rs972295961 Chr.11: 62575560-62575560 on Build GRCh38
rs764135661 Chr.11: 62575617-62575617 on Build GRCh38
rs751730411 Chr.11: 62575536-62575536 on Build GRCh38
rs765249030 Chr.11: 62575513-62575515 on Build GRCh38
rs778057507 Chr.11: 62575529-62575529 on Build GRCh38
rs760433287 Chr.11: 62575518-62575518 on Build GRCh38
rs777606971 Chr.11: 62575533-62575534 on Build GRCh38
rs753600738 Chr.11: 62575521-62575521 on Build GRCh38
rs756938803 Chr.11: 62575544-62575545 on Build GRCh38
rs773698369 Chr.11: 62575431-62575433 on Build GRCh38
rs758529683 Chr.11: 62575526-62575526 on Build GRCh38
rs372977043 Chr.11: 62575506-62575506 on Build GRCh38
rs1040478874 Chr.11: 62575508-62575508 on Build GRCh38
rs965930053 Chr.11: 62575533-62575533 on Build GRCh38
rs147326331 Chr.11: 62575522-62575522 on Build GRCh38
rs139345196 Chr.11: 62575551-62575551 on Build GRCh38
rs774621274 Chr.11: 62575434-62575434 on Build GRCh38
rs766441551 Chr.11: 62575503-62575504 on Build GRCh38
rs751355092 Chr.11: 62575541-62575541 on Build GRCh38
rs191258788 Chr.11: 62575582-62575582 on Build GRCh38
rs757438015 Chr.11: 62575539-62575539 on Build GRCh38

View Full Product Details