Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37231891-37232148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFAF7
Entrez Gene ID
55471
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 7
Gene Aliases
C2orf56, MidA, PRO1853
UniGene
Hs.433466
Gene Chromosome Location
Chr.2: 37231631-37259052 on Build GRCh38
Transcripts
NM_001083946.1 NM_144736.4 XM_005264410.1 XM_005264411.1 XM_011532956.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6467225 c.67A>T p.I23F Chr.2: 37232117-37232117 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs905520157 Chr.2: 37231972-37231972 on Build GRCh38
rs761283905 Chr.2: 37232059-37232059 on Build GRCh38
rs968809275 Chr.2: 37232038-37232038 on Build GRCh38
rs1018797410 Chr.2: 37232044-37232044 on Build GRCh38
rs1020451424 Chr.2: 37232037-37232037 on Build GRCh38
rs1036326270 Chr.2: 37231982-37231982 on Build GRCh38
rs112675974 Chr.2: 37232013-37232013 on Build GRCh38
rs562848214 Chr.2: 37232106-37232106 on Build GRCh38
rs763069033 Chr.2: 37232072-37232072 on Build GRCh38
rs1002524584 Chr.2: 37231986-37231986 on Build GRCh38
rs1046878581 Chr.2: 37231916-37231916 on Build GRCh38
rs777000661 Chr.2: 37232063-37232063 on Build GRCh38
rs764485999 Chr.2: 37232061-37232061 on Build GRCh38
rs2430496 Chr.2: 37232040-37232040 on Build GRCh38
rs538969985 Chr.2: 37231909-37231909 on Build GRCh38
rs750582490 Chr.2: 37232067-37232067 on Build GRCh38
rs200137973 Chr.2: 37232066-37232066 on Build GRCh38
rs150348047 Chr.2: 37231970-37231970 on Build GRCh38
rs149365406 Chr.2: 37232014-37232014 on Build GRCh38
rs904824245 Chr.2: 37231976-37231976 on Build GRCh38
rs554613850 Chr.2: 37231989-37231989 on Build GRCh38
rs556987212 Chr.2: 37231962-37231962 on Build GRCh38
rs530135276 Chr.2: 37231999-37231999 on Build GRCh38
rs536262250 Chr.2: 37231977-37231977 on Build GRCh38
rs17020344 Chr.2: 37232065-37232065 on Build GRCh38
rs892960659 Chr.2: 37231942-37231942 on Build GRCh38
rs766524728 Chr.2: 37232075-37232075 on Build GRCh38
rs1027121293 Chr.2: 37232015-37232015 on Build GRCh38
rs751573166 Chr.2: 37232076-37232076 on Build GRCh38
rs752547992 Chr.2: 37232117-37232117 on Build GRCh38
rs888215668 Chr.2: 37231938-37231938 on Build GRCh38
rs754629398 Chr.2: 37232085-37232089 on Build GRCh38
rs879403451 Chr.2: 37231931-37231931 on Build GRCh38
rs770532499 Chr.2: 37232055-37232055 on Build GRCh38
rs953025828 Chr.2: 37232019-37232019 on Build GRCh38
rs754967979 Chr.2: 37232090-37232090 on Build GRCh38
rs1005303459 Chr.2: 37231949-37231949 on Build GRCh38
rs947151550 Chr.2: 37231969-37231969 on Build GRCh38

View Full Product Details