Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 34767219-34767488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF599
Entrez Gene ID
148103
Gene Name
zinc finger protein 599
Gene Aliases
-
UniGene
Hs.590961
Gene Chromosome Location
Chr.19: 34758074-34773229 on Build GRCh38
Transcripts
NM_001007248.2 XM_017026369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4851088 c.187C>A p.H63N Chr.19: 34767370-34767370 on Build GRCh38
7149165 c.215G>C p.R72T Chr.19: 34767342-34767342 on Build GRCh38
3532353 c.151C>T p.P51S Chr.19: 34767406-34767406 on Build GRCh38
4897460 c.151C>T p.P51S Chr.19: 34767406-34767406 on Build GRCh38
5855453 c.157C>T p.P53S Chr.19: 34767400-34767400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764497564 Chr.19: 34767377-34767377 on Build GRCh38
rs1001444018 Chr.19: 34767329-34767329 on Build GRCh38
rs187901097 Chr.19: 34767442-34767442 on Build GRCh38
rs766631070 Chr.19: 34767383-34767383 on Build GRCh38
rs772462432 Chr.19: 34767291-34767291 on Build GRCh38
rs773616721 Chr.19: 34767324-34767324 on Build GRCh38
rs772996672 Chr.19: 34767373-34767373 on Build GRCh38
rs769386857 Chr.19: 34767448-34767448 on Build GRCh38
rs776405236 Chr.19: 34767439-34767439 on Build GRCh38
rs770508246 Chr.19: 34767461-34767462 on Build GRCh38
rs746615144 Chr.19: 34767297-34767297 on Build GRCh38
rs377243385 Chr.19: 34767292-34767292 on Build GRCh38
rs542079942 Chr.19: 34767437-34767437 on Build GRCh38
rs770628654 Chr.19: 34767301-34767301 on Build GRCh38
rs747255985 Chr.19: 34767349-34767349 on Build GRCh38
rs889430944 Chr.19: 34767307-34767307 on Build GRCh38
rs959714509 Chr.19: 34767380-34767380 on Build GRCh38
rs778053741 Chr.19: 34767432-34767432 on Build GRCh38
rs530217737 Chr.19: 34767320-34767320 on Build GRCh38
rs145532423 Chr.19: 34767328-34767328 on Build GRCh38
rs775172061 Chr.19: 34767455-34767455 on Build GRCh38
rs775618494 Chr.19: 34767337-34767337 on Build GRCh38
rs770484575 Chr.19: 34767438-34767438 on Build GRCh38
rs770991851 Chr.19: 34767361-34767361 on Build GRCh38
rs746665312 Chr.19: 34767424-34767424 on Build GRCh38
rs762812024 Chr.19: 34767354-34767354 on Build GRCh38
rs763941547 Chr.19: 34767359-34767359 on Build GRCh38
rs74834692 Chr.19: 34767319-34767319 on Build GRCh38
rs147712651 Chr.19: 34767410-34767410 on Build GRCh38
rs369901331 Chr.19: 34767413-34767413 on Build GRCh38
rs765840564 Chr.19: 34767461-34767461 on Build GRCh38
rs748487278 Chr.19: 34767268-34767268 on Build GRCh38
rs779530366 Chr.19: 34767265-34767265 on Build GRCh38
rs200678796 Chr.19: 34767418-34767418 on Build GRCh38
rs758924449 Chr.19: 34767421-34767421 on Build GRCh38
rs755375784 Chr.19: 34767262-34767262 on Build GRCh38
rs1010494843 Chr.19: 34767256-34767256 on Build GRCh38
rs753860988 Chr.19: 34767403-34767403 on Build GRCh38
rs900894968 Chr.19: 34767392-34767392 on Build GRCh38

View Full Product Details