Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47264796-47265021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC9
Entrez Gene ID
26093
Gene Name
coiled-coil domain containing 9
Gene Aliases
-
UniGene
Hs.227782
Gene Chromosome Location
Chr.19: 47256474-47271958 on Build GRCh38
Transcripts
NM_015603.2 XM_017026578.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3764755 c.575G>A p.R192Q Chr.19: 47264801-47264801 on Build GRCh38
5364726 c.675delC p.D227fs*26 Chr.19: 47264901-47264901 on Build GRCh38
5031584 c.712G>A p.E238K Chr.19: 47264938-47264938 on Build GRCh38
3749524 c.645G>C p.E215D Chr.19: 47264871-47264871 on Build GRCh38
6501835 c.582C>T p.F194F Chr.19: 47264808-47264808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766811639 Chr.19: 47264921-47264921 on Build GRCh38
rs762121355 Chr.19: 47264910-47264910 on Build GRCh38
rs377072160 Chr.19: 47264863-47264863 on Build GRCh38
rs897671203 Chr.19: 47264895-47264895 on Build GRCh38
rs200381759 Chr.19: 47264973-47264973 on Build GRCh38
rs567723167 Chr.19: 47264882-47264882 on Build GRCh38
rs964976063 Chr.19: 47264972-47264972 on Build GRCh38
rs200234430 Chr.19: 47264904-47264904 on Build GRCh38
rs755816844 Chr.19: 47264883-47264883 on Build GRCh38
rs1010021375 Chr.19: 47264915-47264915 on Build GRCh38
rs763363009 Chr.19: 47264920-47264920 on Build GRCh38
rs202074409 Chr.19: 47264857-47264857 on Build GRCh38
rs529448151 Chr.19: 47264827-47264827 on Build GRCh38
rs201111116 Chr.19: 47264833-47264833 on Build GRCh38
rs201513324 Chr.19: 47264940-47264940 on Build GRCh38
rs988738067 Chr.19: 47264964-47264964 on Build GRCh38
rs773212656 Chr.19: 47264824-47264824 on Build GRCh38
rs754617753 Chr.19: 47264853-47264853 on Build GRCh38
rs534168821 Chr.19: 47264877-47264877 on Build GRCh38
rs993436511 Chr.19: 47264823-47264823 on Build GRCh38
rs765668773 Chr.19: 47264875-47264875 on Build GRCh38
rs558779868 Chr.19: 47264979-47264979 on Build GRCh38
rs753680941 Chr.19: 47264952-47264952 on Build GRCh38
rs569098125 Chr.19: 47264858-47264858 on Build GRCh38
rs752063406 Chr.19: 47264929-47264929 on Build GRCh38
rs946933052 Chr.19: 47264872-47264872 on Build GRCh38
rs776255827 Chr.19: 47264820-47264820 on Build GRCh38
rs773558758 Chr.19: 47264917-47264917 on Build GRCh38
rs778934146 Chr.19: 47264962-47264962 on Build GRCh38
rs202156396 Chr.19: 47264947-47264947 on Build GRCh38
rs777602055 Chr.19: 47264884-47264884 on Build GRCh38
rs770988713 Chr.19: 47264892-47264892 on Build GRCh38
rs965800630 Chr.19: 47264838-47264838 on Build GRCh38
rs150553117 Chr.19: 47264814-47264814 on Build GRCh38
rs757120629 Chr.19: 47264958-47264958 on Build GRCh38
rs200098648 Chr.19: 47264836-47264836 on Build GRCh38
rs780641278 Chr.19: 47264881-47264881 on Build GRCh38
rs747030757 Chr.19: 47264903-47264903 on Build GRCh38
rs749135400 Chr.19: 47264887-47264887 on Build GRCh38
rs768749441 Chr.19: 47264867-47264867 on Build GRCh38
rs777635498 Chr.19: 47264948-47264948 on Build GRCh38
rs750948280 Chr.19: 47264840-47264840 on Build GRCh38
rs968323400 Chr.19: 47264999-47264999 on Build GRCh38
rs758928050 Chr.19: 47264937-47264937 on Build GRCh38
rs200857202 Chr.19: 47264849-47264849 on Build GRCh38
rs2032811 Chr.19: 47264871-47264871 on Build GRCh38
rs988423771 Chr.19: 47264870-47264870 on Build GRCh38
rs764418073 Chr.19: 47264831-47264831 on Build GRCh38
rs769602417 Chr.19: 47264822-47264822 on Build GRCh38
rs759621777 Chr.19: 47264825-47264825 on Build GRCh38
rs374702564 Chr.19: 47264880-47264880 on Build GRCh38
rs746557968 Chr.19: 47264850-47264850 on Build GRCh38
rs968436564 Chr.19: 47264942-47264942 on Build GRCh38
rs202227718 Chr.19: 47264905-47264905 on Build GRCh38
rs762020143 Chr.19: 47264874-47264874 on Build GRCh38
rs754150222 Chr.19: 47264832-47264832 on Build GRCh38
rs766609342 Chr.19: 47264901-47264901 on Build GRCh38
rs752197270 Chr.19: 47264941-47264941 on Build GRCh38
rs926608422 Chr.19: 47264879-47264879 on Build GRCh38
rs538184871 Chr.19: 47264861-47264861 on Build GRCh38
rs747837480 Chr.19: 47264856-47264856 on Build GRCh38
rs760906023 Chr.19: 47264864-47264864 on Build GRCh38
rs757868241 Chr.19: 47264848-47264848 on Build GRCh38
rs760957343 Chr.19: 47264828-47264828 on Build GRCh38
rs760344736 Chr.19: 47264893-47264894 on Build GRCh38
rs779052907 Chr.19: 47264894-47264894 on Build GRCh38
rs554313823 Chr.19: 47264878-47264878 on Build GRCh38
rs377289724 Chr.19: 47264815-47264815 on Build GRCh38
rs201241449 Chr.19: 47264914-47264914 on Build GRCh38
rs780952824 Chr.19: 47264855-47264855 on Build GRCh38
rs748039161 Chr.19: 47264821-47264821 on Build GRCh38

View Full Product Details